Skip to Content
Merck
All Photos(1)

Key Documents

D3415

Sigma-Aldrich

Bisphenol A diglycidyl ether

Bisphenol A diglycidyl ether
1 of 1 reviewers received a sample product or took part in a promotion

viscous liquid, PPARγ inhibitor

Synonym(s):

BADGE, DGEBA, NSC 5022, 2,2-Bis[4-(glycidyloxy)phenyl]propane, 4,4′-Isopropylidenediphenol diglycidyl ether, D.E.R. 332

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H24O4
CAS Number:
Molecular Weight:
340.41
Beilstein:
299026
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Bisphenol A diglycidyl ether,

form

viscous liquid

composition

Epoxide equivalent weight, 172-176

density

1.16 g/mL at 25 °C (lit.)

SMILES string

CC(C)(c1ccc(OCC2CO2)cc1)c3ccc(OCC4CO4)cc3

InChI

1S/C21H24O4/c1-21(2,15-3-7-17(8-4-15)22-11-19-13-24-19)16-5-9-18(10-6-16)23-12-20-14-25-20/h3-10,19-20H,11-14H2,1-2H3

InChI key

LCFVJGUPQDGYKZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bisphenol A diglycidyl ether is an epoxy resin.[1]

Application

Bisphenol A diglycidyl ether has been used:
  • to study its effects on propiomelanocortin (POMC):enhanced green fluorescent protein (EGFP) expression in our transgenic fish[2]
  • to study the curing behavior, kinetics of epoxy, polyamidoamine systems and the influence of the incorporation of Fe3O4 magnetic nanoparticles (MNPs)[3]
  • as a thermosetting resin[4]
  • to determine the optimum composition of the epoxy-polysiloxane blend[5]
  • for the preparation of transparent epoxy-based nanocomposite coatings[6]
  • for the preparation, coating and curing of epoxy-graphene (E/G)[7]

Biochem/physiol Actions

PPARγ inhibitor that blocks rosiglitazone- and insulin-induced adipogenesis.

Features and Benefits

This compound is featured on the Nuclear Receptors (PPARs) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Linkage

Epoxy resin often used in conjunction with DER 732 to form blocks of varying strength and hardness.

Legal Information

D.E.R. is a trademark of The Dow Chemical Company or an affiliated company of Dow

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

507.2 - 514.4 °F - closed cup

Flash Point(C)

264 - 268 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design and Manufacture of Textile Composites (2005)
Enhanced Charpy impact strength of epoxy resin modified with vinyl-terminated polydimethylsiloxane
Vilcakova J, et al.
Journal of Applied Polymer Science, 135(4), 45720-45720 (2018)
Hoa H Le et al.
Endocrinology, 151(12), 5689-5699 (2010-10-12)
Estrogenic endocrine disrupting chemicals (EDCs) constitute a diverse group of man-made chemicals and natural compounds derived from plants and microbial metabolism. Estrogen-like actions are mediated via the nuclear hormone receptor activity of estrogen receptor (ER)α and ERβ and rapid regulation
Enhanced protective properties and UV stability of epoxy/graphene nanocomposite coating on stainless steel.
Alhumade H, et al.
Express Polymer Letters, 10(12) (2016)
H M Wright et al.
The Journal of biological chemistry, 275(3), 1873-1877 (2000-01-15)
While searching for natural ligands for the peroxisome proliferator-activated receptor (PPAR) gamma, we identified a synthetic compound that binds to this receptor. Bisphenol A diglycidyl ether (BADGE) is a ligand for PPARgamma with a K(d(app)) of 100 microM. This compound

Questions

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service