Skip to Content
Merck
All Photos(4)

Documents

135011

Sigma-Aldrich

1,2-Dihydroxybenzene

ReagentPlus®, ≥99%

Synonym(s):

Pyrocatechol, 1,2-Benzenediol, 1,2-Dihydroxybenzene, 2-Hydroxyphenol, Catechol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-(OH)2
CAS Number:
Molecular Weight:
110.11
Beilstein:
471401
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

vapor density

3.8 (vs air)

Quality Level

vapor pressure

1 mmHg ( 75 °C)
10 mmHg ( 118.3 °C)

product line

ReagentPlus®

Assay

≥99%

form

solid

bp

245 °C (lit.)

mp

100-103 °C (lit.)

SMILES string

Oc1ccccc1O

InChI

1S/C6H6O2/c7-5-3-1-2-4-6(5)8/h1-4,7-8H

InChI key

YCIMNLLNPGFGHC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2-Dihydroxybenzene (a phenolic compound) is a useful industrial chemical. It is an ortho isomeric form of dihydroxybenzene. It is commonly referred to as catechol or pyrocatechol. Its standard molar enthalpy of combustion and sublimation at 298.15K have been evaluated by static-bomb calorimetry and microcalorimetry, respectively.

Application

1,2-Dihydroxybenzene may be used as raw material for the industrial synthesis of rubber, dyes, plastics, pharmaceuticals and cosmetics.
1,2-Dihydroxybenzene in combination with amberlite XAD-2, can be used as a chelating resin in the determination of metal ions, using atomic absorption spectrophotometry (AAS).

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Carc. 1B - Eye Dam. 1 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

260.6 °F - closed cup

Flash Point(C)

127 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, characterization and applications of pyrocatechol modified amberlite XAD-2 resin for preconcentration and determination of metal ions in water samples by flame atomic absorption spectrometry (FAAS)
Tewari KP and Singh KA
Talanta, 53(4), 823- 833 (2001)
Catechol suppresses EGF-induced cell transformation by inhibiting ERK2 activity as confirmed by a crystallographic study.
Malakhova M, et al.
Cancer Research, 73(8), 2234-2234 (2013)
Andrew A Horwitz et al.
Cell systems, 1(1), 88-96 (2016-05-03)
CRISPR-Cas genome engineering in yeast has relied on preparation of complex expression plasmids for multiplexed gene knockouts and point mutations. Here we show that co-transformation of a single linearized plasmid with multiple PCR-generated guide RNA (gRNA) and donor DNA cassettes
Thiago C Canevari et al.
The Analyst, 138(1), 315-324 (2012-11-17)
This paper describes the development, characterization and application of an Nb(2)O(5) film formed on the surface of a carbon ceramic material, SiO(2)/C, obtained by a sol-gel method, using the spin-coating technique. The working electrode using this material will be designated
Enthalpies of combustion of 1, 2-dihydroxybenzene and of six alkylsubstituted 1, 2-dihydroxybenzenes.
Da Silva MDMCR, et al.
The Journal of Chemical Thermodynamics, 16(12), 1149-1155 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service