Skip to Content
Merck
All Photos(2)

Documents

430749

Sigma-Aldrich

Pyrocatechol

purified by sublimation, ≥99.5%

Synonym(s):

1,2-Benzenediol, 1,2-Dihydroxybenzene, 2-Hydroxyphenol, Catechol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-(OH)2
CAS Number:
Molecular Weight:
110.11
Beilstein:
471401
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

material

white

Quality Level

vapor density

3.8 (vs air)

vapor pressure

1 mmHg ( 75 °C)
10 mmHg ( 118.3 °C)

Assay

≥99.5%

purified by

sublimation

bp

245 °C (lit.)

mp

100-103 °C (lit.)

SMILES string

Oc1ccccc1O

InChI

1S/C6H6O2/c7-5-3-1-2-4-6(5)8/h1-4,7-8H

InChI key

YCIMNLLNPGFGHC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Pyrocatechol may be used in the synthesis of micro-mesoporous carbon aerogels.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Carc. 1B - Eye Dam. 1 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

260.6 °F - closed cup

Flash Point(C)

127 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Surface characteristics and electrochemical capacitances of carbon aerogels obtained from resorcinol and pyrocatechol using boric and oxalic acids as polymerization catalysts.
Moreno-Castilla C, et al.
Carbon, 49(12), 3808-3819 (2011)
Eagleson M.
Concise Encyclopedia Chemistry, 915-915 (1994)
Adsorbate-induced modification of surface electronic structure: pyrocatechol adsorption on the anatase TiO2 (101) and rutile TiO2 (110) surfaces.
Syres KL, et al.
The Journal of Physical Chemistry C, 116(44), 23515-23525 (2012)
Jacques Poupaert et al.
Current medicinal chemistry, 12(7), 877-885 (2005-04-28)
The 2(3H)-benzoxazolone heterocycle and its bioisosteric surrogates (such as 2(3H)-benzothiazolinone, benzoxazinone, etc.) have received considerable attention from the medicinal chemists owing to their capacity to mimic a phenol or a catechol moiety in a metabolically stable template. These heterocycles and
R D Irons
Journal of toxicology and environmental health, 16(5), 673-678 (1985-01-01)
Occupational exposure to benzene has long been associated with toxicity to the blood and bone marrow, including lymphocytopenia, pancytopenia, aplastic anemia, acute myelogenous leukemia, and possibly lymphoma. A variety of studies have established that benzene itself is not the toxic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service