Skip to Content
Merck
All Photos(1)

Key Documents

11615

Sigma-Aldrich

Boric anhydride

granulated, ≥98.0% (T)

Synonym(s):

Boron trioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
B2O3
CAS Number:
Molecular Weight:
69.62
EC Number:
MDL number:
UNSPSC Code:
12352306
eCl@ss:
38120103
PubChem Substance ID:
NACRES:
NA.21

vapor density

>1 (vs air)

Quality Level

Assay

≥98.0% (T)
98.0-102.0% (titration)

form

solid

quality

granulated

loss

≤3% loss on ignition, 800 °C

mp

450 °C (lit.)

density

2.46 g/mL at 25 °C (lit.)

cation traces

Fe: ≤300 mg/kg

SMILES string

O=BOB=O

InChI

1S/B2O3/c3-1-5-2-4

InChI key

JKWMSGQKBLHBQQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Boric anhydride also referred to as boron trioxide or boron oxide (B2O3) has been used for preparing specimens of Bi4B2O9 ceramics. It has also been used for WO3/B2O3/Al system for investigating the impact of Ar gas pressure on the SHS reaction products.

Biochem/physiol Actions

Boric anhydride is an oxide of boron that shows antimicrobial property along with lower aminoalcohols.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Antimicrobial properties of the products from the reaction of various aminoalcohols and boric anhydride.
S Watanabe
Materials Chemistry and Physics, 19, 191-195 (1988)
Effect of Bi4B2O9 addition on the sintering temperature and microwave dielectric properties of BaO-Nd2O3-4TiO2 ceramics.
Xingyu Chen
Journal of Materials Science: Materials in Electronics, 24, 224-229 (2013)
Influence of Ar Pressure on Product Characteristics of Self-Propagating High-Temperature Synthesis from WO3/B2O3/Al Reactant System.
Tawat Chanadee et al.
Advanced Materials Research, 748, 32-35 (2013)
Yinling Wang et al.
Analytical biochemistry, 385(1), 184-186 (2008-11-27)
Glassy carbon electrode modified with boron oxide nanoparticles supported on multiwall carbon nanotubes was obtained via a facile approach. The as-prepared modified electrode exhibits excellent electrocatalytic activity toward the redox of glucose in pH 7.0 phosphate buffer solution. The electrochemical
Pratik Koirala et al.
The journal of physical chemistry. A, 117(6), 1310-1318 (2012-11-03)
Using density functional theory based calculations, we have systematically studied the equilibrium geometries, relative stabilities, and electronic and magnetic properties of Fe and Mn atoms interacting with a varying number of BO(2) moieties. These clusters are found to exhibit hyperhalogen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service