Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Boric anhydride
Synonym(s):
Boron trioxide
Empirical Formula (Hill Notation):
B
2
O
3
CAS Number:
1303-86-2
Molecular Weight:
69.62
EC Number:
215-125-8
Products
Product Category
salts (2)
bases (1)
Brand
Sigma-Aldrich (3)
Supelco (1)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Reagent Type
Core
Special Grade
Puriss (1)
Puriss P.A. (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
202851
Boric anhydride,
99.999% trace metals basis
Expand
View Pricing
339075
Boric anhydride,
99.98% trace metals basis
Expand
View Pricing
11615
Boric anhydride,
granulated, ≥98.0% (T)
Expand
View Pricing
15678
Boric anhydride,
puriss. p.a., ≥98% (T)
Expand
View Pricing