Skip to Content
Merck
All Photos(1)

Key Documents

09906

Supelco

Cellulose

DFS-0, microcrystalline, suitable for thin layer chromatography (TLC)

Synonym(s):

Cellulose powder, Cotton linters

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41115711
NACRES:
NC.07

Quality Level

form

microcrystalline

technique(s)

thin layer chromatography (TLC): suitable

loss

≤5% loss on drying

InChI

1S/C12H22O11/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-20H,1-2H2/t3?,4?,5?,6?,7?,8?,9?,10-,11?,12+/m1/s1

InChI key

GUBGYTABKSRVRQ-WFVLMXAXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

High purity cellulose powders for partition chromatography.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ginger Brininstool et al.
BMC plant biology, 8, 58-58 (2008-05-20)
The Arabidopsis thaliana CONSTITUTIVE EXPRESSOR OF PATHOGENESIS-RELATED GENES5 (CPR5) gene has been previously implicated in disease resistance, cell proliferation, cell death, and sugar sensing, and encodes a putative membrane protein of unknown biochemical function. Trichome development is also affected in
Jung Min Lee et al.
JAMA otolaryngology-- head & neck surgery, 139(6), 629-635 (2013-06-22)
Bioresorbable fixation systems have been popular for the treatment of facial fractures. However, their mechanical properties are uncertain and complications have been reported. To overcome these problems, we developed a bioresorbable fixation plate using a composite of silk fibroin and
Battle for the barrel.
Robert F Service
Science (New York, N.Y.), 339(6126), 1374-1377 (2013-03-23)
Kornelia Batko et al.
Polimery w medycynie, 43(1), 11-19 (2013-07-03)
The reflection coefficient of the membrane (sigma) is one of the basic parameters of the polymer membrane transport. Classical methods used to determine this parameter require intensive mixing of two solutions separated by a membrane to eliminate the effects of
Ivana Salopek Cubrić et al.
Collegium antropologicum, 37(1), 57-64 (2013-05-24)
Thermophysiological comfort applies to the way in which clothing lets through or retains heat and moisture and helps the body retain heat balance in rest position or at various levels of activities. In this paper, the principles of sensory analysis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service