Skip to Content
Merck
All Photos(3)

Key Documents

S6790

Sigma-Aldrich

Sigmacell Cellulose

Type 101, Highly purified, fibers

Synonym(s):

Cellulose, Cellulose powder, Cotton linters

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.56

type

Type 101

form

fibers

SMILES string

O1[C@H](C(C(C(C1CO)O)O)O)O[C@@H]2C(OC(C(C2O)O)O)CO

InChI

1S/C12H22O11/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-20H,1-2H2/t3?,4?,5?,6?,7?,8?,9?,10-,11?,12+/m1/s1

InChI key

GUBGYTABKSRVRQ-WFVLMXAXSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

High purity cellulose powders for partition chromatography.
Sigmacell cellulose is used in thin layer chromatography (TLC) and cellulose TLC adsorbents. Sigmacell cellulose has been used in studies to inform industrial applications for large-scale purification of enzymes as well as to investigate the cellulase system of Penicillium echinulatum, which has been identified as a potential cellulase producer for bioconversion processes.

Reconstitution

For TLC, simply blend a 15-20% aqueous slurry for about one minute and coat plates; dry at room temperature. Usually does not require activation before use.

Legal Information

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Okako Omadjela et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(44), 17856-17861 (2013-10-16)
Cellulose is a linear extracellular polysaccharide. It is synthesized by membrane-embedded glycosyltransferases that processively polymerize UDP-activated glucose. Polymer synthesis is coupled to membrane translocation through a channel formed by the cellulose synthase. Although eukaryotic cellulose synthases function in macromolecular complexes
Eshchar Mizrachi et al.
The New phytologist, 194(1), 54-62 (2012-04-05)
Fast-growing, short-rotation forest trees, such as Populus and Eucalyptus, produce large amounts of cellulose-rich biomass that could be utilized for bioenergy and biopolymer production. Major obstacles need to be overcome before the deployment of these genera as energy crops, including
Inka Vanwonterghem et al.
The ISME journal, 8(10), 2015-2028 (2014-04-18)
A replicate long-term experiment was conducted using anaerobic digestion (AD) as a model process to determine the relative role of niche and neutral theory on microbial community assembly, and to link community dynamics to system performance. AD is performed by
Nathalie Lavoine et al.
Carbohydrate polymers, 90(2), 735-764 (2012-07-31)
Interest in microfibrillated cellulose (MFC) has been increasing exponentially. During the last decade, this bio-based nanomaterial was essentially used in nanocomposites for its reinforcement property. Its nano-scale dimensions and its ability to form a strong entangled nanoporous network, however, have
Nasrullah Shah et al.
Carbohydrate polymers, 98(2), 1585-1598 (2013-09-24)
Bacterial cellulose (BC) has received substantial interest owing to its unique structural features and impressive physico-mechanical properties. BC has a variety of applications in biomedical fields, including use as biomaterial for artificial skin, artificial blood vessels, vascular grafts, scaffolds for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service