Skip to Content
Merck
All Photos(1)

Documents

08714

Millipore

Methyl Red solution

suitable for microbiology

Synonym(s):

Methyl red indicator solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H15N3O2
CAS Number:
Molecular Weight:
269.30
Beilstein:
1843037
MDL number:
UNSPSC Code:
41171621
PubChem Substance ID:
NACRES:
NA.85

Agency

according to GB 4789.30-2016
according to ISO 22964:2017

Quality Level

product line

BioChemika

shelf life

limited shelf life, expiry date on the label

composition

dist. water, 200 mL
ethanol 95%, 300 mL
methyl red, 0.1 g

technique(s)

microbe id | metabolite detection: suitable

application(s)

clinical testing
environmental
food and beverages

microbiology

suitability

Enterococcus spp.
Escherichia coli
Klebsiella spp.
Proteus spp.
Pseudomonas spp.
Staphylococcus spp.
bacteria

SMILES string

CN(C)c1ccc(cc1)\N=N\c2ccccc2C(O)=O

InChI

1S/C15H15N3O2/c1-18(2)12-9-7-11(8-10-12)16-17-14-6-4-3-5-13(14)15(19)20/h3-10H,1-2H3,(H,19,20)/b17-16+

InChI key

CEQFOVLGLXCDCX-WUKNDPDISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl red solution is an azo dye which turns to red when pH is below 4.4 (yellow pH < 6.2, orange pH 4.4-6.2). Some bacteria utilize glucose to form large amounts of acid with the result that the pH value of the medium falls distinct. Other species produce no or less free acid. This difference can be visualized by using methyl red. This test is used to differentiate enteric bacteria.

Application

Methyl red solution is a pH indicator dye recommended for detection of bacteria producing acid from glucose fermentation such as Enterobacteriaceae in various samples.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

69.8 °F

Flash Point(C)

21 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robin L Stingley et al.
Journal of medical microbiology, 59(Pt 1), 108-114 (2009-09-05)
Reduction of Methyl Red (MR) and Orange II (Or II) by 26 human skin bacterial species was monitored by a rapid spectrophotometric assay. The analysis indicated that skin bacteria, representing the genera Staphylococcus, Corynebacterium, Micrococcus, Dermacoccus and Kocuria, were able
S S Gomare et al.
Journal of applied microbiology, 106(3), 993-1004 (2009-02-04)
To evaluate the potential of Brevibacillus laterosporus MTCC 2298 for the decolourization of different textile azo dyes including methyl red, mechanism of biotransformation and the toxicity of products. Brevibacillus laterosporus showed decolourization of thirteen different azo dyes including methyl red.
X D Zhang et al.
Journal of hazardous materials, 170(2-3), 883-887 (2009-06-09)
An aqueous C.I. Acid Red 2 solution was decolorized by electrolysis using iron as anode. The decolorization mechanism was investigated through experimental observations on the electrochemical behavior of C.I. Acid Red 2 on Pt rotating disk electrode, UV-visible spectra of
Danmeng Shuai et al.
Environmental science & technology, 44(5), 1773-1779 (2010-02-11)
Azo dyes are widespread pollutants and potential cocontaminants for nitrate; we evaluated their effect on catalytic reduction of a suite of oxyanions, diatrizoate, and N-nitrosodimethylamine (NDMA). The azo dye methyl orange significantly enhanced (less than or equal to a factor
Sofia Babanova et al.
Journal of bioscience and bioengineering, 112(4), 379-387 (2011-07-26)
Soluble artificial mediators are often applied to enhance the electron transfer from living cells to an anode in microbial fuel cells. Recently, we have demonstrated that the Candida melibiosica 2491 yeast strain possesses electrogenic properties and can be used as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service