Skip to Content
Merck
All Photos(1)

Key Documents

29333

Millipore

Barritt′s Reagent A

suitable for microbiology

Synonym(s):

Naphtol, ethanolic solution, VP1 Reagent, Voges-Proskauer Test with Barritt’s Reagent A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

Quality Level

Agency

according to ISO 11290-1:2017
according to ISO 11290-2:2017
according to ISO 22964:2017

product line

BioChemika

shelf life

limited shelf life, expiry date on the label

composition

absolute ethanol, 100 mL
α-napthol (1-naphthol), 5.0 g

technique(s)

microbe id | metabolite detection: suitable

application(s)

clinical testing
environmental
food and beverages
pharmaceutical

microbiology

suitability

Enterobacter spp.
Klebsiella spp.
bacteria

Application

This reagent is used in Voges-Proskauer test for detection of acetoin production by bacterial cultures along with Barritt Reagent B.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F

Flash Point(C)

14 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service