Skip to Content
Merck
All Photos(1)

Key Documents

03000590

Ginsenoside Re

primary reference standard

Synonym(s):

(3β,6α,12β)-20-(β-D-Glucopyranosyloxy)-3,12-dihydroxydammar-24-en-6-yl 2-O-(6-deoxy-α-L-mannopyranosyl)-β-D-glucopyranoside, Chikusetsusaponin IVc, Ginsenoside B2, NSC 308877, Panaxoside Re, Sanchinoside Re

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H82O18
CAS Number:
Molecular Weight:
947.15
UNSPSC Code:
85151701
NACRES:
NA.24

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

application(s)

food and beverages

SMILES string

CC1O[C@@H](O[C@@H]2[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]2O[C@H]3C[C@]4(C)[C@H](C[C@@H](O)[C@@H]5[C@H](CC[C@@]45C)[C@](C)(CC\C=C(/C)C)O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O)[C@@]7(C)CC[C@H](O)C(C)(C)[C@H]37)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C48H82O18/c1-21(2)11-10-14-48(9,66-42-38(60)35(57)32(54)26(19-49)63-42)23-12-16-46(7)30(23)24(51)17-28-45(6)15-13-29(52)44(4,5)40(45)25(18-47(28,46)8)62-43-39(36(58)33(55)27(20-50)64-43)65-41-37(59)34(56)31(53)22(3)61-41/h11,22-43,49-60H,10,12-20H2,1-9H3/t22-,23-,24+,25-,26+,27+,28?,29-,30+,31-,32+,33+,34+,35?,36-,37+,38+,39+,40-,41-,42-,43+,45+,46+,47+,48-/m0/s1

InChI key

PWAOOJDMFUQOKB-QVPADXGFSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference Standard in the analysis of herbal medicinal products

Other Notes

This compound is commonly found in plants of the genus: panax

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Souren Paul et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 50(5), 1354-1361 (2012-03-10)
This study was undertaken to evaluate the effect of ginsenoside-Re (Gin-Re) isolated from roots of Panax ginseng on carrageenan-induced paw and TPA-induced skin inflammations in experimental mice. Moreover, to confirm further the anti-inflammatory activities of Gin-Re, LPS-induced macrophage activation model
Hai-Yan Quan et al.
International journal of molecular medicine, 29(1), 73-80 (2011-10-06)
Ginsenoside Re is a protopanaxatriol-type saponin isolated from Panax ginseng berry. Although anti-diabetic and anti-hyperlipidemic effects of Re have been reported by several groups, its mechanism of action is largely unknown until now. Here, we examine anti-diabetic and anti-hyperlipidemic activities
Dan Lu et al.
Journal of ethnopharmacology, 144(3), 656-663 (2012-10-16)
Ginseng has been widely used for hundreds of years in both China and other countries. It is well accepted that the pharmacological effects of ginseng are attributed to ginsenosides. Ginsenoside Re is one of the active ingredients in ginseng. The
Daofeng Qu et al.
Experimental parasitology, 135(2), 234-239 (2013-07-31)
Toxoplasma gondii, the etiological agent of toxoplasmosis, is an obligate intracellular protozoan parasite that infects a variety of mammals including humans. In an attempt to find new antigen-adjuvant combinations that enhance the immunogenicity of antigen candidates for toxoplasma vaccines, we
Kang-Woo Lee et al.
BMC complementary and alternative medicine, 12, 196-196 (2012-10-30)
Microglial activation plays an important role in neurodegenerative diseases by producing several pro-inflammatory enzymes and pro-inflammatory cytokines. Lipopolysaccharide (LPS)-induced inflammation leads to the activation of microglial cells in the central nervous system (CNS) and is associated with the pathological mechanisms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service