Skip to Content
Merck
All Photos(1)

Key Documents

BGALD-RO

Roche

β-Galactose Dehydrogenase

from Pseudomonas fluorescens

Synonym(s):

D-galactose dehydrogenase, D-galactose:NAD+ 1-oxidoreductase

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352204

biological source

Pseudomonas fluorescens

Quality Level

recombinant

expressed in E. coli

form

suspension

specific activity

~5 units/mg protein (At 25 °C with D-galactose as the substrate; standardized with BSA.)

mol wt

64,000 Da

packaging

pkg of 1 mL (10104973001 [1 mg])
pkg of 1 mL (10104981001 [5 mg])

manufacturer/tradename

Roche

technique(s)

activity assay: suitable

color

white

optimum pH

8.4 (0.1 M in Tris buffer)

pH range

4.6-9.2

solubility

water: soluble

suitability

suitable for enzyme test

UniProt accession no.

foreign activity

ADH <0.01%
LDH <0.5%
NADH oxidase <0.1%
galactosidase <0.01%

storage temp.

2-8°C

Related Categories

General description

β-Galactose Dehydrogenase plays a crucial role in galactose metabolism by catalyzing the pyridine-nucleotide-dependent dehydrogenation of β-D-galactopyranose to yield NADH and D-galactono-l,5-1actone. D-galactose dehydrogenase from P. fluorescens is a member of the oxidoreductases family. It is composed of two identical subunits and has a molecular weight of 64,000. Additionally, the enzyme contains two binding sites.

Specificity

Galactose dehydrogenase will oxidize D-galactose [Km = 0.7 mM (30 °C, pH 9.1); relative rate = 1.00], L-arabinose (relative rate = 0.47), D-fucose (relative rate = 1.05), 2-deoxy-D-galactose (relative rate = 0.27), 2-amino-2-deoxy-D-galactose [galactosamine], (relative rate = 0.02; pH 8.6).
Galactose dehydrogenase does not oxidize L-galactose, D-arabinose, D-glucose, L-fucose, D-ribose, D-xylose, D-glucuronic acid, D-galacturonic acid or D-galactose-6-phosphate. The enzyme will use either NAD (Km = 0.24 mM; relative rate = 1.0) or NADP (Km = 2.3 mM; relative rate = 0.32).

Application

β-Galactose Dehydrogenase has been used as a reporter enzyme in neuraminidase (NA) kinetic parameters for the natural substrates 6′-Sialyl-N-acetyllactosamine (6SLN) and 3′-Sialyl-N-acetyllactosamine (3SLN).

Quality

Contaminants: <0.01% ADH, <0.01% ?-galactosidase, <0.1% NADH oxidase, <0.5% LDH

Sequence

Galactose dehydrogenase from Pseudomonas fluorescens (MW 64,000 D) is a dimer with subunits of equal size (subunit MW = 32,000 D).

Unit Definition

One unit galactose dehydrogenase will oxidize 1 mol of β-D-galactose in one minute at +25 °C and pH 8.6.
The QC assay produces 1 mol of NADH per mol of galactose oxidized.

Physical form

Suspension in 3.2 M ammonium sulfate solution, 1 mM EDTA, pH approximately 6

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biophysical measurement of the balance of influenza a hemagglutinin and neuraminidase activities
Benton D, et al.
The Journal of biological chemistry, 290(10), 6516-6521 (2015)
Recombinant D-galactose dehydrogenase partitioning in aqueous two-phase systems: effect of pH and concentration of PEG and ammonium sulfate
Kianmehr A, et al.
Bioresources and bioprocessing, 1.0 (2014)
[29] d-Galactose dehydrogenase from Pseudomonas fluorescens
E. Maier and G. Kurz
Methods in Enzymology, 89.0, 176-181 (1982)
Biophysical Measurement of the Balance of Influenza A Hemagglutinin and Neuraminidase Activities*
Benton, et al.
The Journal of Biological Chemistry, 290, 6516-6521 (2015)
Modelling severe Staphylococcus aureus sepsis in conscious pigs: are implications for animal welfare justified?
Olsen, et al.
BMC Research Notes, 9 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service