Skip to Content
Merck
All Photos(1)

Documents

10745731001

Roche

β-Galactosidase

EIA grade

Synonym(s):

β galactosidase, β-galactosidase

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352204

biological source

Escherichia coli

Quality Level

Assay

(single peak, HPLC)

form

lyophilized

specific activity

750-950 U/mg

mol wt

540.000  kDa

packaging

pkg of 25 mg (approx. 100 mg lyophilizate)

manufacturer/tradename

Roche

technique(s)

enzyme immunoassay: suitable

color

white

pH

7.3-7.7

solubility

water: 20 mg/mL

λmax

405

suitability

suitable for immunoassay

NCBI accession no.

UniProt accession no.

application(s)

life science and biopharma

shipped in

dry ice

storage temp.

−20°C (−15°C to −25°C)

Gene Information

Escherichia coli ... lacZ(945006)

General description

β-Galactosidase, EIA grade, is a lyophilizate from E. coli overproducer, consisting of enzyme protein, phosphate buffer, and sucrose. Substances which could interfere with the derivatization of NH2 or SH groups (e.g., 2-mercaptoethanol, ammonium salts, primary amines etc.) have been removed.
The GLB1 (β-Galactosidase) gene is mapped to human chromosome 3p22.3. The encoded protein belongs to glycosyl hydrolase 35 family and is localized to lysosomes.

Application

β-Galactosidase has been used as a standard to determine the absolute quantitation of LacZ protein.
β-Galactosidase is used for labeling enzyme immunoassay techniques. β-Galactosidase can be coupled to other proteins via its SH-groups. The reconstituted solution can be used directly for conjugation without prior dialysis or gel permeation chromatography.

Biochem/physiol Actions

β-Galactosidase catalyzes the hydrolysis of terminal β-galactose residue of ganglioside substrates, such as glycoproteins, sphingolipids, and keratan sulfate and other glycoconjugates. This enzyme is associated with the mechanism of cell senescence and carcinogenesis. Mutations in the gene result in gangliosidosis, an autosomal recessive disorder, characterized with defective lysosomal storage due to accumulation of substrates. β-Galactosidase deficiency also causes Morquio B syndrome indicating skeletal abnormalities, short stature and increased excretion of keratan sulfate in urine.

Quality

Purity: single peak (HPLC)

Sequence

Free Thiol Groups
The non-denatured, enzymatically-active preparation contains > 12 SH groups per molecule which are not involved in disulfide bridges and are freely accessible to coupling reagents in aqueous media (as assayed with Ellman′s reagent at +37 °C, acc. to Habeeb, 1972).

Absence of Enzyme Aggregates
The preparation contains < 3% dimers (HPLC, area %) and essentially no higher oligomers.

Physical form

Lyophilizate, stabilized with phosphate buffer and sucrose. Note: Contains at least 12 free SH-groups/enzyme molecule; 5 mg approximately 20 mg lyophilizate; 25 mg approximately 100 mg lyophilizate.

Storage and Stability

Store at -15–-25 °C. (sealed under nitrogen)

Analysis Note

Specific activity: approximately 750 - 950 U/mg enzyme protein ≅ approximately 150 - 250 U/mg lyophilizate (+37°C, 2-nitrophenyl-β-D-galactoside); approximately 250-400 U/mg enzyme protein ≅ approximately 60-100 U/mg lyophilizate (+37°C, 4-nitrophenyl-β-D-galactoside).

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Systemic AAV9 gene transfer in adult GM1 gangliosidosis mice reduces lysosomal storage in CNS and extends lifespan.
Weismann C M, et al.
Human Molecular Genetics, 24(15), 4353-4364 (2015)
Hideyuki Yaginuma et al.
Lab on a chip, 22(10), 2001-2010 (2022-04-29)
Digital assays using microreactors fabricated on solid substrates are useful for carrying out sensitive assays of infectious diseases and other biological tests. However, sealing of the microchambers using fluid oil is difficult for non-experts, and thus hinders the widespread use
Association of the GLB1 rs4678680 genetic variant with risk of HBV-related hepatocellular carcinoma.
Wang W T, et al.
Oncotarget, 7(35), 56501-56501 (2016)
Overflow metabolism in Escherichia coli results from efficient proteome allocation.
Basan M, et al.
Nature, 528(7580), 99-99 (2015)
Sasha B Ebrahimi et al.
Nature protocols, 17(2), 327-357 (2022-01-19)
The efficient transfection of functional proteins into cells can serve as a means for regulating cellular processes toward solving fundamental challenges in biology and medicine. However, the use of proteins as effective intracellular agents is hindered by their low cellular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service