11214667001
Roche
Anti-Digoxigenin, Fab fragments
from sheep
Synonym(s):
anti-digoxigenin, digoxigenin
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
Recommended Products
biological source
sheep
Quality Level
conjugate
unconjugated
antibody form
affinity purified immunoglobulin
antibody product type
primary antibodies
clone
polyclonal
form
lyophilized
packaging
pkg of 1 mg
manufacturer/tradename
Roche
technique(s)
ELISA: 2-4 μg/mL
immunohistochemistry: 0.5-1 μg/mL
western blot: 0.5-2 μg/mL
isotype
IgG1
storage temp.
2-8°C
General description
Anti-Digoxigenin, Fab fragments are Fab fragments from polyclonal anti-digoxigenin antibodies. Anti-Digoxigenin Fab fragments are preferentially used for secondary detection using a light chain specific anti-sheep conjugate.
Digoxigenin is a hapten, useful in labeling nucleic acids and in detection systems.
Specificity
The polyclonal antibody from sheep is specific to digoxigenin and digoxin and shows no cross-reactivity with other steroids, such as human estrogens and androgens.
Application
Anti-Digoxigenin Fab fragments are preferentially used for secondary detection using a light chain specific anti-sheep conjugate.
Use Anti-Digoxigenin, Fab fragments for the detection of digoxigenin-labeled compounds using:
Use Anti-Digoxigenin, Fab fragments for the detection of digoxigenin-labeled compounds using:
- ELISA
- Immunohistocytochemistry
- In situ hybridization
- Western blot
- Flow cell construction.
Features and Benefits
Specific to digoxigenin and digoxin.
Sequence
The number of amino groups in the Fab fragments is not known, because polyclonal antibodies are not sequenced. But as a rule of thumb full length antibodies contain about 80 amino groups on average.
Preparation Note
Working concentration: Working concentration of antibody depends on application and substrate. The following concentrations should be taken as a guideline:
Storage conditions (working solution): Diluted antibody is stable at 2 to 8 °C for 12 hours.
Always prepare freshly!
- ELISA: 2 to 4 μg/ml
- Immunohistocytochemistry: 0.5 to 1 μg/ml
- In situ hybridization: 0.2 to 0.4 μg/ml
- Western blot: 0.5 to 2 μg/ml
Storage conditions (working solution): Diluted antibody is stable at 2 to 8 °C for 12 hours.
Always prepare freshly!
Reconstitution
Add 1 ml PBS to a final concentration of 1 mg/ml. Centrifuge reconstituted antibody for 5 minutes at 10,000 rpm prior to each use, and pipet the necessary amount carefully from the surface.
Analysis Note
Preparation: After immunization with digoxigenin, sheep IgGs are purified by ion exchange chromatography. Specific IgGs are isolated by immunosorption. Fab fragments obtained after papain digestion are purified by gel filtration and lyophilized.
Other Notes
For life science research only. Not for use in diagnostic procedures.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
does not flash
Flash Point(C)
does not flash
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Action of thyrocalcitonin on Paget's osteodystrophy
Bompani R, et al.
Giornale di gerontologia, 18(8), 720-734 (1970)
Véronique Van De Bor et al.
Development (Cambridge, England), 148(4) (2021-02-03)
Basement membranes (BM) are extracellular matrices assembled into complex and highly organized networks essential for organ morphogenesis and function. However, little is known about the tissue origin of BM components and their dynamics in vivo Here, we unravel the assembly
Annealing helicase HARP closes RPA-stabilized DNA bubbles non-processively
Burnham DR, et al.
Nucleic Acids Research, 45(8), 4687-4695 (2017)
Daniel J Jackson et al.
Autophagy, 10(3), 408-415 (2013-12-18)
Representatives of all major metazoan lineages form biominerals. The molecular mechanisms that underlie this widespread and evolutionarily ancient ability are gradually being revealed for some lineages. However, until a wider range of metazoan biomineralization strategies are understood, the true diversity
J Brooks Crickard et al.
The EMBO journal, 39(20), e105705-e105705 (2020-08-14)
Rad54 and Rdh54 are closely related ATP-dependent motor proteins that participate in homologous recombination (HR). During HR, these enzymes functionally interact with the Rad51 presynaptic complex (PSC). Despite their importance, we know little about how they are organized within the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service