Skip to Content
Merck
All Photos(1)

Key Documents

11333089001

Roche

Anti-Digoxigenin

from sheep

Synonym(s):

anti-digoxigenin, digoxigenin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Pricing and availability is not currently available.

biological source

sheep

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized

packaging

pkg of 200 μg

manufacturer/tradename

Roche

isotype

IgG

storage temp.

2-8°C

General description

Digoxigenin is a hapten which is used in labeling of nucleic acids and in detection systems.[1]
Probes labeled with digoxigenin has greater sensitivity equivalent to that of radioactive probes. It allows faster detection, is less hazardous and has an increased shelf life.[2]

Specificity

The polyclonal antibody from sheep is specific to digoxigenin and digoxin and shows no cross-reactivity with other steroids, such as human estrogens and androgens.

Application

Anti-Digoxigenin has been used in DNA tethering.[3] It has been used to attach DNA molecule to the glass surface of the flow cell.[4]
Use Anti-Digoxigenin antibody for the detection of digoxigenin-labeled compounds using:
  • ELISA
  • Immunohistocytochemistry[5]
  • In situ hybridization[6]
  • Western blot

Biochem/physiol Actions

In the presence of Na+, Mg2+ and ATP, digoxigenin inhibits sodium pumps.[7]

Preparation Note

Working concentration: Working concentration of antibody depends on application and substrate. The following concentrations should be taken as a guideline:
  • ELISA: for coating: 2 to 4 μg/ml
  • Immunohistocytochemistry: 0.5 to 2 μg/ml
  • In situ hybridization: 0.2 to 0.4 μg/ml
  • Western blot: 0.5 to 2 μg/ml

Working solution: For coating applications: PBS (phosphate buffered saline), pH 7.4
After immunization with digoxigenin, sheep IgG was purified by ion-exchange chromatography, and the specific IgG was isolated by immunosorption.

Reconstitution

Add 1 ml PBS to a final concentration of 200 μg/ml.

Analysis Note

No cross reactivity with other steroids, such as human estrogens e.g., estradiol or androgens e.g., testosterone.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Methods in Molecular Biology (2011)
Affinity Labeling of a Sulfhydryl Group in the Cardiacglycoside Receptor Site of Na+/K+?ATPase by N?Hydroxysuccinimidyl Derivatives of Digoxigenin.
Antolovic R, et al.
European Journal of Biochemistry, 227(1?2), 61-67 (1995)
Juliette Salvaing et al.
PloS one, 7(5), e38156-e38156 (2012-06-14)
In the mouse zygote, DNA methylation patterns are heavily modified, and differ between the maternal and paternal pronucleus. Demethylation of the paternal genome has been described as an active and replication-independent process, although the mechanisms responsible for it remain elusive.
Chan Chen et al.
Aging, 12(14), 14490-14505 (2020-07-22)
Cardiac arrest (CA) is the leading cause of death around the world. Survivors after CA and cardiopulmonary resuscitation (CPR) develop moderate to severe cognitive impairment up to 60% at 3 months. Accumulating evidence demonstrated that long non-coding RNAs (lncRNAs) played
Nonradioactive labeling of probe with digoxigenin by polymerase chain reaction.
Lion T and Haas O A
Analytical Biochemistry, 188(2), 335-337 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service