Skip to Content
Merck
All Photos(1)

Key Documents

11175041910

Roche

DIG Nucleic Acid Detection Kit

greener alternative

sufficient for 40 blots (10 cm x 10 cm each), kit of 1 (5 components), suitable for hybridization, suitable for Northern blotting

Synonym(s):

DIG system

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116134

usage

sufficient for 40 blots (10 cm x 10 cm each)

Quality Level

packaging

kit of 1 (5 components)

manufacturer/tradename

Roche

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

Northern blotting: suitable
Southern blotting: suitable
hybridization: suitable

greener alternative category

storage temp.

−20°C

General description

DIG Nucleic Acid Detection Kit is used for the detection of digoxigenin-labeled nucleic acids by enzyme immunoassay and enzyme-catalyzed color reaction. DIG-labeled nucleic acids are detected after hybridization in an enzyme-linked immunoassay with a highly specific anti-DIG-AP (alkaline phosphatase) antibody conjugate and the color substrates NBT (nitroblue tetrazolium) and BCIP (5-bromo-4-chloro-3-indolyl-phosphate).
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is designed as a safer chemical.  The DIG System was established as a sensitive and cost-effective alternative to radioactivity for the labeling and detection of nucleic acids. There are many available publications that prove the versatility of the DIG System, so use of radio-labeling is no longer the only option for labeling of DNA for hybridization.

Application

DIG nucleic acid detection kit has been used in the confirmation of genetic manipulation.
The DIG Nucleic Acid Detection Kit contains all essential reagents for the detection of membrane-blotted, digoxigenin-labeled nucleic acid hybrids. It can be used in a variety of hybridization techniques:
  • Southern blots
  • Northern blots
  • Other nucleic acid blotting applications
  • In situ hybridization applications
The kit contains all essential reagents for the detection of membrane-blotted, digoxigenin-labeled nucleic acid hybrids. It can be used in a variety of hybridization techniques:
  • Southern blots
  • Northern blots
  • Other nucleic acid blotting applications
  • In situ hybridization applications

Features and Benefits

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is designed as a safer chemical. The DIG System was established as a sensitive and cost-effective alternative to using radioactivity for the labeling and detection of nucleic acids. There are many available publications that prove the versatility of the DIG System, so use of radio-labeling is no longer the only option for labeling of DNA for hybridization.

Packaging

1 kit containing 5 components.

Principle

DIG-labeled nucleic acids are detected after hybridization in an enzyme-linked immunoassay with a highly specific anti-DIG-AP antibody conjugate and the color substrates NBT and BCIP.

Preparation Note

Working concentration: Flow cytometry: 2 μg/ml (0.2 μg/100 μl/106 cells); Immunohistocytochemistry: 6 μg/ml
Working concentration of conjugate depends on application and substrate.
Storage conditions (working solution):
  • Anti-Digoxigenin-AP Conjugate (vial 3): 2 to 8 °C, it is stable for 12 months at this temperature
  • NBT/BCIP (vial 4): 2 to 8 °C, stable
    Note: During shipment of the kit on dry ice, a precipitate may occur which is dissolved by briefly warming to 37 °C
  • The blocking reagent (vial 5) is stable for 36 months and can be stored dry at 4 to 8 °C.
  • Autoclaved stock solution can be stored for several days to a week either unopened at 15 to 25 °C or at 4 to 8 °C after opening. Alternatively, it can be stored in aliquots at -15 to -25 °C for up to 6 months.

Kit Components Only

Product No.
Description

  • DIG-labeled Control DNA 5 µg/ml

  • DNA Dilution Buffer, [50 μg/ml] fish sperm DNA

  • Anti-Digoxigenin-AP Conjugate antibody 750 U/ml

  • NBT/BCIP, 50x stock solution 50x concentrated

  • Blocking Reagent

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

does not flashNot applicable

Flash Point(C)

does not flashNot applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiromasa Funato et al.
Nature, 539(7629), 378-383 (2016-11-05)
Sleep is conserved from invertebrates to vertebrates, and is tightly regulated in a homeostatic manner. The molecular and cellular mechanisms that determine the amount of rapid eye movement sleep (REMS) and non-REMS (NREMS) remain unknown. Here we identify two dominant
Han Yang et al.
Nature communications, 13(1), 485-485 (2022-01-27)
Nitrogen (N), one of the most important plant nutrients, plays crucial roles in multiple plant developmental processes. Spikelets are the primary sink tissues during reproductive growth, and N deficiency can cause floral abortion. However, the roles of N nutrition in
Zheyong Xue et al.
Nature communications, 9(1), 604-604 (2018-02-11)
In flowering plants, the pollen coat protects the released male germ cells from desiccation and damage during pollination. However, we know little about the mechanism by which the chemical composition of the pollen coat prevents dehydration of pollen grains. Here
Zhanwu Yang et al.
Plant biotechnology journal, 20(1), 103-115 (2021-09-07)
Legume-rhizobia symbiosis enables biological nitrogen fixation to improve crop production for sustainable agriculture. Small heat shock proteins (sHSPs) are involved in multiple environmental stresses and plant development processes. However, the role of sHSPs in nodule development in soybean remains largely
The role of Aspartyl aminopeptidase (Ape4) in Cryptococcus neoformans virulence and authophagy
Gontijo FA, et al.
PLoS ONE (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service