Skip to Content
Merck
All Photos(1)

Key Documents

11175025910

Roche

DIG RNA Labeling Kit (SP6/T7)

greener alternative

sufficient for 2 x 10 labeling reactions, kit of 1 (12 components), suitable for hybridization, suitable for Southern blotting

Synonym(s):

DIG system, rna labeling

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105500
Pricing and availability is not currently available.

usage

sufficient for 2 x 10 labeling reactions

Quality Level

packaging

kit of 1 (12 components)

manufacturer/tradename

Roche

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

Northern blotting: suitable
Southern blotting: suitable
hybridization: suitable

greener alternative category

storage temp.

−20°C

General description

Assay Time: 145minutes
Sample Materials
  • Linearized plasmid DNA
  • PCR product
Principle
The DIG RNA Labeling Kit produces DIG-labeled, single-stranded RNA probes of known length. Either SP6 or T7 RNA polymerase transcribes these probes in vitro from template DNA (in the presence of digoxigenin-UTP).
RNA Labeling by in vitro Transcription
The DNA to be transcribed is cloned into the polylinker site of appropriate transcription vectors (e.g., pSPT 18 or 19), which contain promoters for SP6 and T7 RNA polymerases. Adjacent template DNA is linearized at a suitable site and the RNA polymerases are used to produce "run off" transcripts. DIG-UTP is incorporated into the transcript. Every 20 to 25th nucleotide of the newly synthesized RNA is a DIG-UTP. Since the nucleotide concentration does not become limiting in the standard transcription reaction, this reaction can generate large amounts of labeled RNA.
Kit for the labeling of RNA with digoxigenin-UTP by in vitro transcription with SP6 and T7 polymerases. By this method, single-stranded RNA probes of known length are produced, which can be used in a variety of hybridization techniques.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is designed as a safer chemical.  The DIG System was established as a sensitive and cost-effective alternative to using radioactivity for the labeling and detection of nucleic acids. There are many available publications that prove the versatility of the DIG System, so use of radio-labeling is no longer the only option for labeling of DNA for hybridization.

Specificity

Sensitivity and Specificity
DIG-labeled RNA probes can detect single-copy genes in as little as 1 μg of mammalian DNA under the following assay conditions: The hybridization mix contains 20 to 100 ng labeled probe/ ml, and the bound probe is detected with anti-DIG-AP and visualized with the chemiluminescent substrate CDP-Star.
Heat inactivation: Stop the reaction by adding 2 μl 0.2 M EDTA (pH 8.0).

Application

For RNA labeling with DIG-11-UTP by in vitro transcription with SP6 and T7 RNA Polymerases. DIG-labeled "run off" transcripts are synthesized with high efficiency and can be used in a variety of hybridization techniques:
  • Northern blots[1]
  • Southern blots
  • In situ hybridizations[2][3][4]
  • Plaque or colony lifts
  • RNase protection experiments
Due to highly specific and sensitive detection systems, DIG-labeled probes can be used for single-copy gene detection in 1μg total human DNA.
Note: Since the linkage between DIG and UTP is resistant to alkali, DIG-labeled RNA can be fragmented by alkaline treatment. Slightly reducing the size of the DIG-labeled RNA probe may make it more suitable for certain applications in in situ hybridization.

Packaging

1 kit containing 12 components.

Quality

Test principle: The DNA template to be transcribed is cloned into the polylinker site of appropriate transcription vectors, which contain promoters for SP6 and/or T7 RNA Polymerases. After linearization at a suitable site, RNA is transcribed in the presence of DIG-UTP. Under standard conditions, approximately 10μg of full-length DIG-labeled RNA is transcribed from 1μg template.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Kit Components Only

Product No.
Description

  • pSPT18 DNA 0.25 mg/ml

  • pSPT19 DNA 0.25 mg/ml

  • Control DNA 1, pSPT18-Neo, cleaved with Pvu II 0.25 mg/ml

  • Control DNA 2, pSPT19-Neo, cleaved with Pvu II 0.25 mg/ml

  • DIG-labeled Control RNA, DIG-labeled "antisense" neo RNA 100 ng/µl

  • Unlabeled Control RNA, neo poly (A) "sense" RNA 200 µg/ml

  • NTP Labeling Mixture 10x concentrated

  • Transcription Buffer 10x concentrated

  • DNase I, RNase-free 10 U/µl

  • Protector RNase Inhibitor 20 U/µl

  • SP6 RNA Polymerase 20 U/µl

  • T7 RNA Polymerase 20 U/µl

See All (12)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Karlgren et al.
Journal of visualized experiments : JoVE, (26)(26), doi:10-doi:10 (2009-04-21)
The high-throughput expression analysis technologies available today give scientists an overflow of expression profiles but their resolution in terms of tissue specific expression is limited because of problems in dissecting individual tissues. Expression data needs to be confirmed and complemented
Yu Liang et al.
Molecular medicine reports, 5(4), 1087-1091 (2012-01-17)
Schistosoma japonicum (S. japonicum) is an extremely harmful pathogen, which infects humans and causes severe public health problems. To date, no effective therapeutic drugs for this pathogen are available. In this study, we designed and constructed three hammerhead ribozymes targeting
Nathan Luebbering et al.
PloS one, 8(10), e76775-e76775 (2013-10-23)
The DYRKs (dual-specificity tyrosine phosphorylation-regulated kinases) are a conserved family of protein kinases that are associated with a number of neurological disorders, but whose biological targets are poorly understood. Drosophila encodes three Dyrks: minibrain/Dyrk1A, DmDyrk2, and DmDyrk3. Here we describe
Parvaneh Alizadeh et al.
Experimental eye research, 83(3), 679-687 (2006-05-11)
Cystatin C is the major inhibitor of the cysteine cathepsins. Polymorphisms in the cystatin C gene have recently been associated with the risk of developing Age-related Macular Degeneration (AMD). Oxidative stress is also thought to play a key role in
Peng Li et al.
Development (Cambridge, England), 138(9), 1795-1805 (2011-03-25)
Secreted factors from the epicardium are believed to be important in directing heart ventricular cardiomyocyte proliferation and morphogenesis, although the specific factors involved have not been identified or characterized adequately. We found that IGF2 is the most prominent mitogen made

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service