Sign In to View Organizational & Contract Pricing.
Select a Size
Pricing and availability is not currently available.
About This Item
UNSPSC Code:
41105339
NACRES:
NA.54
Skip To
Product Name
Nylon Membranes, positively charged, roll W × L 0.3 m × 3 m, sheet W × L 10 cm × 15 cm, sheet W × L 20 cm × 30 cm
packaging
pkg of 10 sheets (11209272001[20 x 30 cm])
pkg of 20 sheets (11209299001[10 x 15 cm])
manufacturer/tradename
Roche
roll W × L
0.3 m × 3 m
sheet W × L
10 cm × 15 cm
20 cm × 30 cm
storage temp.
20-25°C
Quality Level
1 of 4
This Item | 11699075001 | 15356 | 58067 |
|---|---|---|---|
| roll W × L 0.3 m × 3 m | roll W × L - | roll W × L - | roll W × L - |
| packaging pkg of 10 sheets (11209272001[20 x 30 cm]) | packaging pkg of 50 discs (82 mm diameter) | packaging - | packaging pkg of 50 ea |
| manufacturer/tradename Roche | manufacturer/tradename Roche | manufacturer/tradename - | manufacturer/tradename - |
| sheet W × L 10 cm × 15 cm | sheet W × L - | sheet W × L - | sheet W × L - |
| storage temp. 20-25°C | storage temp. 20-25°C | storage temp. room temp | storage temp. - |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level - |
Analysis Note
Function tested for homogenous DNA detection in Southern blot procedures using the DIG System.
Application
- The physical and chemical properties of the nylon membranes make them specifically useful as a matrix for hybridization with nonradioactively (e.g., digoxigenin or biotin) and radioactively (e.g., [32P], [35S], an [3H])-labeled DNA, RNA, or oligonucleotide probes: Southern blots[1]
- Northern blots[2]
- Dot blots
Features and Benefits
Microporous nylon 66 membrane on a polyester support, carrying positively charged quaternary ammonium groups.
Contents: Membranes
Contents: Membranes
General description
Microporous nylon 66 membrane on a polyester support, carrying positively charged (pH 3 - 10) quaternary ammonium groups.
Surface Properties: Pore size of 0.45μm
Surface Properties: Pore size of 0.45μm
Other Notes
For life science research only. Not for use in diagnostic procedures.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Majid Kheirollahi et al.
Medical oncology (Northwood, London, England), 28(3), 864-870 (2010-04-08)
Telomeres at the ends of human chromosomes consist of tandem hexametric (TTAGGG)n repeats, which protect them from degradation. At each cycle of cell division, most normal somatic cells lose approximately 50-100 bp of the terminal telomeric repeat DNA. Precise prediction of
Araceli Nora García et al.
BMC plant biology, 14, 248-248 (2014-09-18)
The production of antimicrobial peptides is a common defense strategy of living cells against a wide range of pathogens. Plant snakin peptides inhibit bacterial and fungal growth at extremely low concentrations. However, little is known of their molecular and ecological
Dervla T Isaac et al.
Infection and immunity, 81(2), 411-420 (2012-11-28)
Histoplasma capsulatum is a fungal respiratory pathogen that survives and replicates within the phagolysosome of macrophages. The molecular factors it utilizes to subvert macrophage antimicrobial defenses are largely unknown. Although the ability of H. capsulatum to prevent acidification of the
Baoyan Bai et al.
Nucleus (Austin, Tex.), 4(4), 315-325 (2013-06-21)
CRM1 is an export factor that together with its adaptor NMD3 transports numerous cargo molecules from the nucleus to cytoplasm through the nuclear pore. Previous studies have suggested that CRM1 and NMD3 are detected in the nucleolus. However, their localization
Jes-Niels Boeckel et al.
Proceedings of the National Academy of Sciences of the United States of America, 108(8), 3276-3281 (2011-02-09)
JmjC domain-containing proteins play a crucial role in the control of gene expression by acting as protein hydroxylases or demethylases, thereby controlling histone methylation or splicing. Here, we demonstrate that silencing of Jumonji domain-containing protein 6 (Jmjd6) impairs angiogenic functions
Related Content
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service

