Skip to Content
Merck
All Photos(1)

Key Documents

11685619910

Roche

Fluorescein RNA Labeling Mix

solution, suitable for Northern blotting, suitable for Southern blotting, suitable for hybridization

Synonym(s):

RNA labeling

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105500
Pricing and availability is not currently available.

form

solution

Quality Level

usage

sufficient for 20 reactions

packaging

pkg of 40 μL

manufacturer/tradename

Roche

technique(s)

Northern blotting: suitable
Southern blotting: suitable
hybridization: suitable

storage temp.

−20°C

General description

Fluorescein RNA Labeling Mix is a convenient nucleotide mixture for the labeling of RNA with fluorescein-12-uridine triphosphate (UTP).

Specificity

Heat inactivation: Stop the reaction by adding 2μl 0.2M EDTA (pH 8.0).

Application

Fluorescein-labeled RNA has been used in a variety of hybridization techniques such as:
  • in situ hybridization to tissues[1]
  • Southern blots
  • northern blots
  • plaque or colony lifts
  • RNase protection experiments

Quality

The mix is function tested in combination with the DIG RNA Labeling Kit.

Principle

Fluorescein-labeled, single-stranded RNA probes of defined length are generated by in vitro transcription. Fluorescein-12-UTP is incorporated by SP6, T7, and T3 RNA polymerases at approximately every 20 to 25th nucleotide of the transcript under standard conditions. The Fluorescein RNA Labeling Mix is specifically designed for use with the RNA polymerases SP6, T7, and T3, which are supplied with an optimized transcription buffer.

Preparation Note

10x concentrated NTP labeling mixture: 10mM ATP, 10mM CTP, 10mM GTP, 6.5mM UTP, 3.5mM Fluorescein-12-UTP, pH 7.5 (+20°C).

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhimin Jiang et al.
Nature microbiology, 6(7), 932-945 (2021-05-15)
The retinoic acid-inducible gene I (RIG-I) receptor senses cytoplasmic viral RNA and activates type I interferons (IFN-I) and downstream antiviral immune responses. How RIG-I binds to viral RNA and how its activation is regulated remains unclear. Here, using IFI16 knockout
Yasufumi Hayano et al.
Science advances, 7(29) (2021-07-16)
The most prominent structural hallmark of the mammalian neocortical circuitry is the layer-based organization of specific cell types and synaptic inputs. Accordingly, cortical inhibitory interneurons (INs), which shape local network activity, exhibit subtype-specific laminar specificity of synaptic outputs. However, the
Lucia Caceres et al.
Developmental dynamics : an official publication of the American Association of Anatomists, 248(12), 1243-1256 (2019-10-01)
Familial exudative vitreoretinopathy (FEVR) is a rare congenital disorder characterized by a lack of blood vessel growth to the periphery of the retina with secondary fibrovascular proliferation at the vascular-avascular junction. These structurally abnormal vessels cause leakage and hemorrhage, while
Anke J Roelofs et al.
Annals of the rheumatic diseases, 79(12), 1625-1634 (2020-09-24)
Osteophytes are highly prevalent in osteoarthritis (OA) and are associated with pain and functional disability. These pathological outgrowths of cartilage and bone typically form at the junction of articular cartilage, periosteum and synovium. The aim of this study was to
Paul A Dawson et al.
Biology of reproduction, 87(2), 43-43 (2012-06-08)
Sulfate is important for mammalian growth and development. During pregnancy, maternal circulating sulfate levels increase by 2-fold, enhancing sulfate availability to the fetus. We used quantitative real-time PCR to determine sulfate transporter mRNA levels during mouse gestation in three tissues:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service