Skip to Content
Merck
All Photos(1)

Key Documents

ZTC18M

Millipore

ZipTip® Pipette Tips

C18 resin, micro, bed volume 0.2 μL, tip volume 10 μL

Synonym(s):

ZipTip Pipette Tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
eCl@ss:
32033590
NACRES:
NB.24

material

polypropylene

Quality Level

sterility

non-sterile

manufacturer/tradename

ZipTip®

parameter

4-70  °C temperature

technique(s)

analytical sample preparation: suitable
fractionation: suitable
protein purification: suitable (concentration)

bed volume

0.2 μL

thickness

15 μm

tip I.D.

3.375 mm

tip L

31 mm

tip O.D.

5.8 mm

tip volume

10 μL

pore size

200 Å pore size

pH

1.5-13.5
2-12(for 24 hour exposure)

capacity

2.0 μg binding capacity(typically; when used with saturating amounts of analyte)

shipped in

ambient

Application

ZipTip® with 0.2 µLC18 resin is suitable for mass spectrometry sample preparation and protein concentration.

Features and Benefits

  • One-step desalting, concentration, and purification
  • Allows fractionation of complex samples
  • Ideal for peptides, proteins, nucleic acids, and more
  • Maximum recovery without dead volume
  • Eliminates time-consuming chromatography

Other Notes

Automation Compatible: Yes

Legal Information

ZIPTIP is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Mark A Jackson et al.
Frontiers in plant science, 10, 602-602 (2019-06-04)
The backbone cyclic and disulfide bridged sunflower trypsin inhibitor-1 (SFTI-1) peptide is a proven effective scaffold for a range of peptide therapeutics. For production at laboratory scale, solid phase peptide synthesis techniques are widely used, but these synthetic approaches are
Sha Liu et al.
Journal of proteome research, 21(7), 1589-1602 (2022-06-18)
Aberrant protein N-glycosylation is a cancer hallmark, which has great potential for cancer detection. However, large-scale and in-depth analysis of N-glycosylation remains challenging because of its high heterogeneity, complexity, and low abundance. Human saliva is an attractive diagnostic body fluid
Choo Hock Tan et al.
Comparative biochemistry and physiology. Part D, Genomics & proteomics, 43, 101006-101006 (2022-06-20)
The Red-headed Krait (Bungarus flaviceps) is a medically important venomous snake species in Southeast Asia, while there is no specific antivenom available for its envenoming. This study investigated the venom composition through a decomplexation proteomic approach, and examined the immunoreactivity
Luis Ariel Espinosa et al.
Analytical and bioanalytical chemistry, 413(30), 7559-7585 (2021-11-06)
Subunit vaccines based on the receptor-binding domain (RBD) of the spike protein of SARS-CoV-2 provide one of the most promising strategies to fight the COVID-19 pandemic. The detailed characterization of the protein primary structure by mass spectrometry (MS) is mandatory
Chen-Chung Liao et al.
Biology, 11(2) (2022-02-26)
Atherosclerosis is the preliminary cause of coronary artery disease, one of the diseases that account for the largest number of fatal mortalities. Physical activity is an effective strategy to restrain atherosclerosis from deterioration. Evidence indicated that changes in the proteomic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service