Skip to Content
Merck
All Photos(4)

Key Documents

A6141

Sigma-Aldrich

Ammonium bicarbonate

ReagentPlus®, ≥99.0%

Synonym(s):

Ammonium hydrogen carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4HCO3
CAS Number:
Molecular Weight:
79.06
Beilstein:
4329606
EC Number:
MDL number:
UNSPSC Code:
12352300
eCl@ss:
38020216
PubChem Substance ID:
NACRES:
NA.21

vapor density

2.7 (vs air)

Quality Level

vapor pressure

513 hPa ( 50 °C)
67.1 hPa ( 20 °C)

product line

ReagentPlus®

Assay

≥99.0%

form

powder or crystals

pH

7-8.5 (25 °C, 79.1 g/L)

SMILES string

N.OC(O)=O

InChI

1S/CH2O3.H3N/c2-1(3)4;/h(H2,2,3,4);1H3

InChI key

ATRRKUHOCOJYRX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium bicarbonate is an inorganic salt commonly used in the food industry. It can be generated by the interaction of gaseous carbon dioxide and aqueous ammonia. The addition of ammonium bicarbonate to microbial fuel cells (MFCs) has been reported to improve its functioning by altering the porosity and pore distribution of catalyst layers. A recent study reports the use of bubble column evaporator (BCE) based analysis of its thermal degradation in aqueous solution.

Application

Ammonium bicarbonate (AMBIC) was used in the following studies:
  • To derivatize histone proteins from human monocyte-derived macrophages (MDM) by propionylation.
  • Destaining of silver-stained proteins in polyacrylamide gel bands during the peptide mass profiling using a mass spectrometer.
  • Digestion of proteins isolated from Xenopus egg extracts in coomassie blue-stained gel bands during the analysis of microtubule-binding proteins.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Burdock GA.
Encyclopedia of Food and Color Additives, 1, 131-131 (1997)
Yao-Fa Lin et al.
Neural regeneration research, 14(12), 2183-2191 (2019-08-10)
Wallerian degeneration and nerve regeneration after injury are complex processes involving many genes, proteins and cytokines. After different peripheral nerve injuries the regeneration rate can differ. Whether this is caused by differential expression of genes and proteins during Wallerian degeneration
Using ammonium bicarbonate as pore former in activated carbon catalyst layer to enhance performance of air cathode microbial fuel cell.
Li D, et al.
Journal of Power Sources, 272, 909-914 (2014)
Pawel Olszowy et al.
Proteome science, 13, 24-24 (2015-09-29)
Histones and their post-translational modifications impact cellular function by acting as key regulators in the maintenance and remodeling of chromatin, thus affecting transcription regulation either positively (activation) or negatively (repression). In this study we describe a comprehensive, bottom-up proteomics approach
Vincent Gache et al.
Methods in molecular medicine, 137, 29-43 (2007-12-19)
Microtubule-binding proteins are conveniently divided into two large groups: MAPs (microtubule-associated proteins), which can stabilize, anchor, and/or nucleate microtubules, and motors, which use the energy of ATP hydrolysis for a variety of functions, including microtubule network organization and cargo transportation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service