Skip to Content
Merck
All Photos(1)

Key Documents

WBULS0500

Millipore

Immobilon ECL Ultra Western HRP Substrate

Western blotting, HRP

Synonym(s):

chemiluminescent detection substrate, detection substrate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116010
eCl@ss:
42029053
NACRES:
NA.32

Quality Level

technique(s)

western blot: suitable

detection method

chemiluminescent

shipped in

ambient

General description

Immobilon® ECL Ultra Western HRP Substrate is an ultra-sensitive reagent with long signal duration for the detection of horseradish peroxidase (HRP)-labeled antibodies on western blots. The intense signal created permits sensitive detection at the mid-to-low femtogram range using x-ray film or digital imaging systems.

Application

"Immobilon® ECL Ultra substrate is compatible with both PVDF andnitrocellulose blotting membranes, as well as commonly used buffers and blocking reagents.

Sufficient for 5000 cm²"
Western blotting, HRP

Components

Solution A- 250 mL

Solution B- 250 mL

Storage and Stability

Store at 2–8 °C

Legal Information

Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service