Skip to Content
Merck
All Photos(1)

Documents

IPVH85R

Millipore

Immobilon®-P Membrane

1 roll, 8.5 cm x 10 m, 0.45 µm pore size, Hydrophobic PVDF Transfer Membrane for western blotting.

Synonym(s):

Immobilon® NOW, Immobilon® narrow roll, Western blotting membrane, blotting membrane, transfer membrane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105339
eCl@ss:
32031613
NACRES:
NB.23

product name

Immobilon®-P Membrane, PVDF, 0.45 m, 8.5 cm x 10 m roll, 0.45 um ( 0.45 micron filter ) pore size Hydrophobic immobilon-P PVDF Transfer Membrane for western blotting using Chemiluminescence, Chromogenic or Radioactive detection methods. Comes in an 8.5 cm x 10 m roll.

material

PVDF membrane
plain filter
white filter

Quality Level

feature

hydrophobic

manufacturer/tradename

Immobilon®

technique(s)

dot blot: suitable
western blot: suitable

filter L × W

8.5 cm × 10 m

pore size

0.45 μm pore size

capacity

160 μg/cm2 adsorption capacity (insulin)
215 μg/cm2 adsorption capacity (BSA)
294 μg/cm2 adsorption capacity (goat IgG)

compatibility

for use with Amido black
for use with CPTS
for use with Colloidal gold
for use with Coomassie brilliant blue
for use with India ink
for use with Ponceau-S red
for use with Sypro<TMSYMBOL></TMSYMBOL> ruby
for use with Toluidine blue
for use with Transillumination

detection method

chemiluminescent
colorimetric
fluorometric
radioactive

shipped in

ambient

General description

Immobilon®-P transfer membrane is a hydrophobic polyvinylidene fluoride (PVDF) microporous membrane with a nominal pore size of 0.45 micron (μm). It is used for the transfer of proteins from a variety of gel matrices. Immobilon®-P transfer membrane is recommended for most western blotting, especially proteins greater than 20 kDa. This membrane displays improved handling characteristics and staining capabilities, a higher signal-to-noise ratio for enhanced sensitivities, and improved solvent resistance, when compared to nitrocellulose membrane.
Primary Binding Mechanisms: Electrostatic, hydrophobic,
Filter Code: IPVH

Application

Immobilon®-P Membrane, PVDF, 0.45 μm, 8.5 cm x 10 m roll has been used in western blot analysis and co-immunoprecipitation.

Linkage

Replaces: INCP00010

Legal Information

Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chunzhi Li et al.
Investigative ophthalmology & visual science, 61(10), 9-9 (2020-08-07)
Retinoblastoma (RB) is the most common primary intraocular tumor in children. Chemoresistance is the major obstacle for treatment of these tumors. This study aims to determine whether or not downregulating microRNA-222 (miR-222) could serve as a potential therapeutic target for
Wei Cai et al.
Gene, 736, 144386-144386 (2020-01-25)
Neuropathic pain, caused by damage to the nerve system, is one of the most challenging diseases in the world. Moreover, the etiology remains unclear. Circular RNAs (circRNAs) have been revealed to participate in various biological progress, including neuropathic pain. However
microRNA-222-Mediated VHL Downregulation Facilitates Retinoblastoma Chemoresistance by Increasing HIF1? Expression.
Chunzhi Li, et al.
Investigative Ophthalmology & Visual Science (2020)
Jeremiah Tsyporin et al.
Cell reports, 35(12), 109269-109269 (2021-06-24)
Projection neuron subtype identities in the cerebral cortex are established by expressing pan-cortical and subtype-specific effector genes that execute terminal differentiation programs bestowing neurons with a glutamatergic neuron phenotype and subtype-specific morphology, physiology, and axonal projections. Whether pan-cortical glutamatergic and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service