Skip to Content
Merck
All Photos(1)

Key Documents

UC0E05

Sigma-Aldrich

UCOE® Intermediate Vector Set

Synonym(s):

UCOE Mammalian Protein Expression Vectors, UCOE Mammalian Protein Expression Plasmids, Ubiquitous Chromatin Opening Element, UCOE05

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106621
eCl@ss:
34360190
NACRES:
NA.51

shipped in

dry ice

Quality Level

General description

The Ubiquitous Chromatin Opening Element (UCOE) technology offers significant advances in recombinant mammalian protein expression. Unlike traditional vectors, UCOE-containing vectors have the capacity to ease the process of isolating cell line clones that express high-levels of recombinant proteins. UCOE sequences promote reproducible and stable high level expression of a linked gene by altering chromatin structure to a transcriptionally permissive state that negates epigenetic-mediated (e.g. DNA methylation) silencing.

CET 1019 contains the 3 kb mouse Rps3 UCOE and the 1.5 kb fragment of human HNRPA2B1-CBX3 UCOE.
CET 1310 contains the 8 kb fragment of human HNRPA2B1-CBX3 UCOE.
CET 1316 contains the 4 kb fragment of human HNRPA2B1-CBX3 UCOE.

References
Antoniou, M., et al.. (2003) Transgenes Encompassing Dual Promoter CpG Islands from the Human TBP and HNRPA2B1 Loci are Resistant to Heterochromatin-mediated Silencing. Genomics, 82: 269-279.
Antoniou, M., et al. (2010) A Ubiquitous Chromatin Opening Element (UCOE) Confers Resistance to DNA Methylation-mediated Silencing of Lentiviral Vectors. Mol Ther, 18: 1640-1649.

THIS PRODUCT IS ONLY AVAILABLE FOR SALE TO ACADEMIC INSTITUTIONS OR NOT-FOR-PROFIT ENTITIES FOR USE UNDER THIS PRODUCT LABEL LICENSE. FOR INFORMATION ON COMMERCIAL LICENSING OF THE PATENTED UCOE TECHNOLOGY, PLEASE CONTACT [email protected].

Components

1) 10ug CET 1019 (3 kb Rps3 and 1.5kb HNRPA2B1-CBX3) Vector (CS221300)

2) 10ug CET 1310 (8 kb HNRPA2B1-CBX3) Vector (CS221301)

3) 10ug CET 1316 (4 kb HNRPA2B1-CBX3) Vector (CS221302)

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

UCOE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Antoniou et al.
Genomics, 82(3), 269-279 (2003-08-09)
The genetic elements that are responsible for establishing a transcriptionally competent, open chromatin structure at a region of the genome that consists only of ubiquitously expressed, housekeeping genes are currently unknown. We demonstrate for the first time through functional analysis
Fang Zhang et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 18(9), 1640-1649 (2010-07-01)
DNA methylation may restrict the activity of gene transfer vectors due to inadvertent silencing. In P19 embryonic carcinoma cells in vitro, we found that transgene expression regulated by the SFFV LTR and EF1 alpha promoter declined rapidly within 16 days

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service