Skip to Content
Merck

Skip To

SEGPU0545

Stericup® E-GP Funnel-less Sterile Vacuum Filtration System

greener alternative

pore size 0.22 μm, polyethersulfone membrane, capacity 500 mL, 45 mm thread, pack of 12 ea

Synonym(s):

Stericup® E-GP Eco-friendly Filter Unit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NB.24
UNSPSC Code:
41104914
Pore size:
0.22 μm pore size
Filter capacity:
500 mL
Packaging:
pack of 12 ea
Material:
polyethersulfone membrane, polystyrene bottle, polystyrene funnel(s), polystyrene housing
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

material

polyethersulfone membrane, polystyrene bottle, polystyrene funnel(s), polystyrene housing

product line

Stericup®

feature

hydrophilic

packaging

pack of 12 ea

greener alternative product characteristics

Waste Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

parameter

max. temp. at 45 °C

description

2.4 mL (hold-up volume)

technique(s)

sterile filtration: suitable

H

188 mm

capacity

500 mL

diam.

9.3 cm

filter capacity

500 mL

filter diam.

73 mm

filtration area

40 cm2

matrix

Express PLUS

pore size

0.22 μm pore size

fitting

inlet thread(s), 45 mm thread

greener alternative category

shipped in

ambient

storage temp.

no temp limit

General description

We are committed to bringing you Greener Alternative Products, which belong to one of the four categories of greener alternatives . This product is a Design for Sustainability (DfS) developed product. Click here to view its DfS scorecard.
Work with ease and filter with confidence with Stericup® and Steritop® Filtration Systems.

The Stericup® E and Steritop® E sterile filters are designed as a Greener Alternative for vacuum sterile filtration that significantly reduces plastic and packaging.

This Stericup® E is a 45 mm thread collar which uses a 0.22 μm pore size polyethersulfone (PES) membrane filter unit combined with a 500 mL receiver flask and cap for processing and storage. The 45 mm thread directly connects to wider neck media bottles (such as Gibco® or glass bottles).

Use only glass or plastic bottles designed for vacuum applications.

Application

For Research Use Only
Stericup® and Steritop® systems are designed to reduce foaming and protein denaturation while maximizing flow and can be used for sterile vacuum filtration of aqueous solutions such as tissue culture media and biological fluids.
  • Tissue culture media +/- additives
  • Buffers
  • Biological solutions

Features and Benefits

  • The filter collar disconnects from the receiver bottle with just a quarter turn—reducing the likelihood of spillage and making it easier to manipulate on the bench or under the hood.
  • Bold product identification on the filter collar plainly displays membrane characteristics, including composition and pore size, for rapid, application-appropriate product selection.
  • Click seal cap is flanged and modified to enable a firm grip in wet or dry conditions—and a tactile stop confirms secure closure. Worry less about the possibility of spilling or contamination of sterilized contents.
  • Frosted, ample writing surface on the receiver bottle and lighter cap color facilitate clear labelling to improve legibility and reduce unreadable smears.
  • Click seal cap rests on the side to avoid risk of contamination.
  • Receiver bottles are stackable to save storage space.

Legal Information

EXPRESS PLUS is a trademark of Merck KGaA, Darmstadt, Germany
STERITOP is a registered trademark of Merck KGaA, Darmstadt, Germany
Stericup is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SEGPU0538SEGPU1138SEGPT0038
pore size

0.22 μm pore size

pore size

0.22 μm pore size

pore size

0.22 μm pore size

pore size

0.22 μm pore size

material

polyethersulfone membrane, polystyrene funnel(s), polystyrene bottle, polystyrene housing

material

polyethersulfone membrane, polystyrene bottle, polystyrene funnel(s), polystyrene housing

material

polyethersulfone membrane, polystyrene bottle, polystyrene funnel(s), polystyrene housing

material

polyethersulfone membrane, polystyrene bottle, polystyrene funnel(s), polystyrene housing

filter capacity

500 mL

filter capacity

500 mL

filter capacity

1000 mL

filter capacity

150 mL , 150 mL × 1000 mL

packaging

pack of 12 ea

packaging

pack of 12 ea

packaging

pack of 12 ea

packaging

pack of 12 ea

fitting

inlet thread(s), 45 mm thread

fitting

inlet thread(s), 38 mm (1 1/2 in.) thread

fitting

inlet thread(s), 38 mm (1 1/2 in.) thread

fitting

inlet thread(s), 38 mm (1 1/2 in.) thread

feature

hydrophilic

feature

hydrophilic

feature

hydrophilic

feature

hydrophilic


Still not finding the right product?




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
SEGPU054504061841074243

Questions

1–10 of 15 Questions  
  1. What Bacterial Endotoxins Test is used?

    1 answer
    1. The test method used complies with USP 85 and EP 2.6.14 to ensure the absence of endotoxin in our products.

      Helpful?

  2. What tubing can be used with Stericup® and Steritop® vacuum filters?

    What tubing can be used with Stericup® and Steritop® vacuum filters?

    1 answer
    1. Stericup® and Steritop® filters are fitted with a 1/4” tube adapter. We recommend 3/16 in. (4.8 mm) ID silicone tubing to fit this tube adapter: https://www.sigmaaldrich.com/product/mm/xx7100004

      Helpful?

  3. Can we filter oil-type liquids with Stericup® filters?

    Can we filter oil-type liquids with Stericup® filters?

    1 answer
    1. Likely not, due to pore size/possible fouling/chemical compatibility. Refer to our guide, "Chemical compatibility of filter components": https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/302/855/membrane-chemical-compatibility-ds2532en-ms.pdf

      Helpful?

  4. What is the chemical compatibility of the various Stericup® and Steritop® filters?

    What is the chemical compatibility of the various Stericup® and Steritop® filters?

    1 answer
    1. Helpful?

  5. The User Guide says “Use only glass or plastic bottles designed for vacuum applications”. Can you specify which bottles are compatible?

    1 answer
    1. Steritop® Quick Release bottle top units require a vacuum-rated receiver bottle with either a GL33 or GL45 mm closure thread as specified. Steritop® E bottle top units require a vacuum-rated receiver bottle with a GL45 mm closure thread. Bottles no larger than 2 L should be used.
      Glass bottles, GL33:
      • Wheaton media bottles (glass), 500 mL (Product Z364886)
      • Wheaton media bottles (glass), 1 L (Product Z364894)
      Glass bottles, GL45
      • Corning® glass media storage bottle, 125 mL (Product CLS1399125)
      • Corning® glass media storage bottle, 250 mL (Product CLS1399250)
      • Corning® glass media storage bottle, 500 mL (Product CLS1399500)
      • Corning® glass media storage bottle, 1 L (Product CLS13991L)
      • Pyrex® round media storage bottle, 500 mL (Product CLS1395500)
      • Pyrex® round media storage bottle, 1 L (Product CLS13951L)
      • KIMBLE® KIMAX® Media bottle, 250 mL (Product DWK14395250)
      • KIMBLE® KIMAX® Media bottle, 500 mL (Product DWK14395500)
      • KIMBLE® KIMAX® Media bottle, 1 L (Product DWK143951000)
      Plastic (pre-sterile) bottles, GL45:
      • Stericup® Quick Release Receiver flask, 250 mL (Product S200B02RE)
      • Stericup® Quick Release Receiver flask, 500 mL (Product S200B05RE)
      • Stericup® Quick Release Receiver flask, 1000 mL (Product S200B10RE)

      Helpful?

  6. What is the expiration date of Stericup® and Steritop® filters?

    1 answer
    1. Typically 3 years from sterilization.

      Helpful?

  7. Are Stericup® and Steritop® filters GMP-certified?

    1 answer
    1. No, Stericup® and Steritop® devices are not GMP-approved.

      Helpful?

  8. Can Stericup® and Steritop® filters be reused?

    1 answer
    1. All Stericup® and Steritop® filters were developed for single use, disposable processing.

      Helpful?

  9. What is the maximum vacuum pressure for filtration?

    1 answer
    1. 700 mm Hg (27.5” Hg, 0.9 bar) differential vacuum at 25 °C (77 °F)

      Helpful?

  10. Can Stericup® bottles be placed inside freezers for long-term storage of media?

    1 answer
    1. You can successfully freeze and store many aqueous solutions (such as culture media) in Stericup® bottles at temperatures up to -20 °C (-4 °F). It is strongly recommended that you run a sample stability trial under your actual storage conditions prior to using Stericup® bottles for frozen storage.

      Helpful?

1–10 of 15 Questions  

Reviews

No rating value

Active Filters