Skip to Content
Merck
All Photos(1)

Key Documents

SCR029

Sigma-Aldrich

Cryopreserved Mouse Cortical Neural Stem Cells

The Cryopreserved Mouse Cortical Neural Stem Cells provides ready-to use primary neural stem cells isolated from the cortices of embryonic day 15-18 (E15-E18) C57/BL6 mice.

Synonym(s):

Cortical Stem Cells, Mouse Neural Stem Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106509
eCl@ss:
32011203
NACRES:
NA.81

biological source

mouse

Quality Level

manufacturer/tradename

Chemicon®

technique(s)

cell culture | stem cell: suitable

input

sample type neural stem cell(s)

shipped in

liquid nitrogen

General description

Millipore′s Cryopreserved Mouse Cortical Neural Stem Cells provides ready-to use primary neural stem cells isolated from the cortices of embryonic day 15-18 (E15-E18) C57/BL6 mice. These primary neural stem cells may be used for a variety of research applications including drug development, for studies of neurotoxicity, neurogenesis, electrophysiology, neurotransmitter and receptor functions and CNS diseases and disorders.

We recommend that Millipore′s Cryopreserved Mouse Cortical Neural Stem Cells (Catalog No. SCR029) be used in conjunction with the Neural Stem Cell Marker Characterization Kit (Catalog No. SCR019) and differentiation assays that demonstrate multipotentiality of the starting cell population.

For Research Use Only; not for use in diagnostic procedure.

Cell Line Description

Neural Stem Cells

Application

Research Category
Stem Cell Research
The Cryopreserved Mouse Cortical Neural Stem Cells provides ready-to use primary neural stem cells isolated from the cortices of embryonic day 15-18 (E15-E18) C57/BL6 mice.

Packaging

1 x 106 cells

Components

1x10e6 viable Mouse Cortical Neural Stem Cells: (Part No. 2004115) derived from mixed gender C57/BL6 embryonic day 15-18 mice, cryopreserved. Store in liquid nitrogen.

Storage and Stability

When stored at the recommended storage conditions (liquid nitrogen), mouse cortical neural stem cells are stable up to the expiration date. Do not expose to elevated temperatures. Discard any remaining reagents after the expiration date. We recommend that the cells be used within ten passages.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
cOmplete is a trademark of Roche

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brian Sims et al.
Brain research, 1321, 88-95 (2010-01-28)
Erythropoietin (Epo) has been used for many years in neonates for the treatment of anemia of prematurity. Epo has also been proposed for treatment of neonatal brain injury, as mounting evidence suggests neuroprotective properties for Epo. However, Epo's neuroprotective mechanism
Mathew Blurton-Jones et al.
Stem cell research & therapy, 5(2), 46-46 (2014-07-16)
Short-term neural stem cell (NSC) transplantation improves cognition in Alzheimer's disease (AD) transgenic mice by enhancing endogenous synaptic connectivity. However, this approach has no effect on the underlying beta-amyloid (Aβ) and neurofibrillary tangle pathology. Long term efficacy of cell based
Cameron D Morley et al.
Advanced materials (Deerfield Beach, Fla.), 35(44), e2304212-e2304212 (2023-09-01)
Injectable hydrogels are increasingly explored for the delivery of cells to tissue. These materials exhibit both liquid-like properties, protecting cells from mechanical stress during injection, and solid-like properties, providing a stable 3D engraftment niche. Many strategies for modulating injectable hydrogels
B Thaci et al.
Cancer gene therapy, 21(1), 38-44 (2014-01-18)
Myeloid-derived suppressor cells (MDSCs) accumulate in the glioma microenvironment during tumor progression and promote immunosuppression. Interleukin-12 (IL-12) immunogene therapy can alter MDSCs toward an antigen-presenting cell phenotype and these mature cells can have a central role in antigen presentation. It
Karen L Ring et al.
Cell stem cell, 11(1), 100-109 (2012-06-12)
The generation of induced pluripotent stem cells (iPSCs) and induced neuronal cells (iNCs) from somatic cells provides new avenues for basic research and potential transplantation therapies for neurological diseases. However, clinical applications must consider the risk of tumor formation by iPSCs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service