Skip to Content
Merck
All Photos(8)

Key Documents

XTG360-RO

Roche

X-tremeGENE 360 Transfection Reagent

Universal polymer reagent for delivering DNA, siRNA, miRNA and CRISPR/RNP to many cell lines

Synonym(s):

transfection reagent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106502
NACRES:
NA.55

grade

for molecular biology

form

liquid (aqueous solution)

packaging

pkg of 0.4 mL (08724105001)
pkg of 1.0 mL (08724121001)
pkg of 5 × 1.0 mL (08724156001)

manufacturer/tradename

Roche

technique(s)

transfection: suitable

storage temp.

−20°C

General description

X-tremeGENE 360 Transfection Reagent is an innovative reagent that forms a complex with DNA or RNA and efficiently delivers the nucleic acids into animal or insect cells. With improved utility, X-tremeGENE 360 delivers high efficiency transfection in more cell types with low cytotoxicity.

Application

X-tremeGENE 360 Transfection Reagent is a high performance transfection reagent, free of animal-derived components.
  • Designed to transfect a broad range of eukaryotic cells, many cell lines not transfected well by other reagents, and hard-to-transfect cell lines, such as asK-562 and HepG2.
  • Can be successfully used in a variety of applications, such as gene expression analysis using transiently transfected cells, generating of stable cell lines, expression of siRNA for gene knockdown studies, and CRISPR gene-editing tools.
  • Produces minimal cytotoxicity or changes in morphology when adequate numbers of cells are transfected, eliminating the requirement to change media after adding the transfection complex.
  • Suitable for transient and stable transfection.
  • Functions well in the presence or absence of serum.

Features and Benefits

X-tremeGENE 360 Transfection Reagent is a high-performing, versatile and reliable solution for hard-to-transfect and common cell types. As a proven and reliable reagent, X-tremeGENE 360 allows researchers to avoid guesswork when performing transfection. In addition, X-tremeGENE 360 exhibits:
  • Peak performance in a broad range of cell types
  • Suitable for multiple applications
  • Efficient delivery of plasmid DNA, siRNA/miRNA and CRISPR/Cas9 components

Quality

Each lot of X-tremeGENE 360 Transfection Reagent is carefully tested following established quality procedures to ensure that the product is consistently performing according to specifications. During quality testing, cells are transfected with a reporter gene vector DNA using X-tremeGENE 360 Transfection Reagent (ratio 3:1 μL/μg DNA). Reporter gene activity is monitored via chemiluminescent detection. Using a standard curve, the total amount of recombinant protein is determined per well in order to meet specification.

Physical form

Supplied in 80% ethanol and sterile-filtered through 0.2 μm pore size membrane. Does not contain any ingredients of human or animal origin. Number of Tests: Using the standard procedure, 1 mL of X-tremeGENE 360 Transfection Reagent can be used to perform up to 10,000 transfections in 96-well plates.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Legal Information

X-tremeGENE is a trademark of Roche

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

61.7 °F

Flash Point(C)

16.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Establishment of a Rapid Detection System for ISG20-Dependent SARS-CoV-2 Subreplicon RNA Degradation Induced by Interferon-?
Furutani, et al.
International Journal of Molecular Sciences, 22 (2021)
Structure of the vasopressin hormone?V2 receptor??-arrestin1 ternary complex
Bous. J, et al.
Science Advances, 8 (2022)
Structure?function?pathogenicity analysis of C-terminal myocilin missense variants based on experiments and 3D models
Zhou, et al.
Frontiers in Genetics, 13 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service