SRP3128
NT-3 human
recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Synonym(s):
HGNF, NGF-2, Nerve growth factor-2, Neurotrophic factor, Neurotrophin-3
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.32
Recommended Products
biological source
human
recombinant
expressed in E. coli
Assay
≥98% (HPLC)
≥98% (SDS-PAGE)
form
lyophilized
potency
≤20 ng/mL ED50/EC50
mol wt
13.6 kDa
packaging
pkg of 10 μg
technique(s)
cell culture | mammalian: suitable
impurities
<0.1 EU/μg endotoxin, tested
color
white to off-white
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... NTF3(4908)
General description
NT-3 (neurotrophin 3) is a neurotrophic factor structurally related to β-NGF (nerve growth factor), BDNF (brain-derived neurotrophic factor), and NT-4. These proteins belong to the cysteine-knot family of growth factors that assume stable dimeric structures. NT-3 is expressed by neurons of the central nervous systems and can signal through the trk receptors. Recombinant human NT-3 is a noncovalently linked homodimer, of two 13.6kDa polypeptide monomers (240 total amino acid residues). The amino acid sequences of human, murine and rat NT-3 are identical.
Biochem/physiol Actions
NT-3 (neurotrophin 3) promotes the growth and survival of nerve and glial cells. It is associated with neuroprotection and polymorphisms in the gene are associated with risk to Alzheimer′s disease. It is also linked with attention-deficit/hyperactivity disorder (ADHD) and mood disorders. NT-3 enhances the metastatic proliferation of breast cancer cells in the brain.
Sequence
MYAEHKSHRG EYSVCDSESL WVTDKSSAID IRGHQVTVLG EIKTGNSPVK QYFYETRCKE ARPVKNGCRG IDDKHWNSQC KTSQTYVRAL TSENNKLVGW RWIRIDTSCV CALSRKIGRT
Physical form
Lyophilized with no additives.
Reconstitution
Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
The neurotrophin family of neurotrophic factors: an overview.
Skaper SD
Methods in Molecular Biology, 846, 1-12 (2012)
Neurotrophin-3 modulates breast cancer cells and the microenvironment to promote the growth of breast cancer brain metastasis.
Louie E, et al.
Oncogene, 32, 4064-4077 (2013)
S O Meakin et al.
Trends in neurosciences, 15(9), 323-331 (1992-09-01)
The neurotrophins, of which nerve growth factor (NGF) is the best known example, support the survival and differentiation of chick embryo sensory neurons at extremely low concentrations, 10(-12) M or less. These same neurons display two different classes of neurotrophin
Neurotrophin 3 genotype and emotional adverse effects of osmotic-release oral system methylphenidate (OROS-MPH) in children with attention-deficit/hyperactivity disorder.
Park S, et al.
Journal of Psychopharmacology, 28, 220-226 (2014)
Aliccia Bollig-Fischer et al.
Molecular biomedicine, 2(1), 26-26 (2022-01-11)
The development of targeted therapies that inhibit cancer-driving oncogenes has improved outcomes of patients diagnosed with lung adenocarcinoma (LUAD). In contrast, patients diagnosed with lung squamous cell carcinoma (LUSC) suffer worse survival outcomes and lack effective targeted treatment options. Identification
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service