Skip to Content
Merck

Skip To

MXPESP5CN

Valve Outlet Cap + Needle

Accessory used in On-line sampling and filtration of aqueous liquids

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
41116129
NACRES:
NB.24
eCl@ss:
32160606
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polypropylene cap

Quality Segment

packaging

pkg of 5 numbers

manufacturer/tradename

MicropreSURE®

input

sample type wine
sample type cider
sample type pharmaceutical(s)
sample type bottled water
sample type beverage(s)
sample type food(s)
sample type sport drink
sample type soft drink
sample type dairy
sample type beer

application(s)

bioburden testing
food and beverages
pharmaceutical

shipped in

ambient

General description

Valve Outlet Cap + Needle is an accessory of the MicropreSure® sampler. MicropreSure® sampler is an on-line filtration sampler to simplify in-process microbiological testing. It is a simple, protective design that allows samples to be aseptically processed right at the collection site thereby, reducing contamination risk. The valve outlet cap + needle is used to cap the value opening post usage.

Application

MicropreSure® valve outlet cap + needle is used for on-line sampling and filtration of aqueous liquids such as beer, dairy, bottled water, pharmaceutical products analysis, soft drink, cider, sports drink, and wine for bioburden testing.

Legal Information

MICROPRESURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MXPESP14NMXPESPTCLMXPESP18N
packaging

pkg of 5 numbers

packaging

pkg of 1 ea

packaging

pkg of 1 ea

packaging

pkg of 1 ea

material

polypropylene cap

material

316L stainless steel

material

316L stainless steel

material

316L stainless steel

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

application(s)

bioburden testing
food and beverages
pharmaceutical

application(s)

bioburden testing
food and beverages
pharmaceutical
water monitoring

application(s)

bioburden testing
food and beverages
pharmaceutical
water monitoring

application(s)

bioburden testing
food and beverages
pharmaceutical
water monitoring

input

sample type wine
sample type cider
sample type pharmaceutical(s)
sample type bottled water
sample type beverage(s)
sample type food(s)
sample type sport drink
sample type soft drink
sample type dairy
sample type beer

input

-

input

-

input

-

Quality Level

SP1

Quality Level

EQ1

Quality Level

EQ1

Quality Level

EQ1




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
MXPESP5CN04053252300073

Questions

Reviews

No rating value

Active Filters