Skip to Content
Merck

MXPESP14N

MicropreSure® Sanitary Sampling Valve

1/4" NPT valve for On-line sampling and filtration of aqueous liquids, 316L stainless steel

Synonym(s):

Stainless steel sampling valve

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116129
NACRES:
NB.24
eCl@ss:
32160606
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

material

316L stainless steel

packaging

pkg of 1 ea

manufacturer/tradename

MicropreSURE®

valve size

1/4 in.

application(s)

bioburden testing
food and beverages
pharmaceutical
water monitoring

compatibility

1/4" NPT valve for On-line sampling and filtration of aqueous liquids

shipped in

ambient

General description

MicropreSure® sanitary sampling valve is an accessory of the MicropreSure® sampler. MicropreSure® sampler is an on-line filtration sampler to simplify in-process microbiological testing. It is a simple, protective design that allows samples to be aseptically processed right at the collection site thereby, reducing contamination risk. The Sanitary sampling valve is installed at the sampling port outlet having the tri-clover connection for easy sampling of the liquids.

Application

MicropreSure® sanitary sampling valve is used for on-line sampling and filtration of aqueous liquids such as beer, dairy, bottled water, pharmaceutical products analysis, soft drink, cider, sports drink, and wine for bioburden testing.

Legal Information

MICROPRESURE is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library