Skip to Content
Merck
All Photos(2)

Key Documents

MABS1180

Sigma-Aldrich

Anti-E3 ubiquitin-protein ligase UBR1 Antibody, clone 6H9.1

clone 6H9.1, from mouse

Synonym(s):

EC: 2.3.2.27, N-recognin-1, RING-type E3 ubiquitin transferase UBR1, Ubiquitin-protein ligase E3-alpha-1, Ubiquitin-protein ligase E3-alpha-I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6H9.1, monoclonal

species reactivity

human, rat

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... UBR1(197131)
rat ... Ubr1(499877)

Related Categories

General description

E3 ubiquitin-protein ligase UBR1 (UniProt: Q8IWV7; also known as EC: 2.3.2.27, N-Recognin-1, RING-type E3 ubiquitin transferase UBR1, Ubiquitin-protein ligase E3-alpha-1, Ubiquitin-protein ligase E3-alpha-I) is encoded by the UBR1 gene (Gene ID: 197131) in human. E3 ubiquitin-protein ligase UBR1 is a component of the N-end rule pathway. It recognizes and binds to proteins bearing specific N-terminal residues that are destabilizing according to the N-end rule, leading to their ubiquitination and subsequent degradation. It may also be involved in pancreatic homeostasis. Two isoforms of UBR1 have been described that are produced by alternative splicing. UBR1 is broadly expressed protein with highest levels reported in skeletal muscle, kidneys, and pancreas. UBR1 has two zinc finger domains. The UBR-type domain is localized to amino acids 97 to 168 and it forms a pocket that mediates recognition of type 1 N-degrons. It exhibits preference for arginine in first position and displays poor affinity for histidine. The second zinc finger domain, RING-type, atypical, is localized to amino acids 1098-1201. Mutations in UBR1 gene are causative factor in Johanson-Blizzard syndrome that is characterized by congenital exocrine pancreatic insufficiency, malformations such as nasal wing aplasia, and mental retardation.

Specificity

Clone 6H9.1 detects E3 ubiquitin-protein ligase UBR1 in human and rat cells. It targets an epitope within 101 amino acids from the internal region.

Immunogen

GST/His-tagged recombinant fragment corresponding to 101 amino acids from the internal region of human E3 ubiquitin-protein ligase UBR1.

Application

Anti-E3 ubiquitin-protein ligase UBR1, clone 6H9.1, Cat. No. MABS1180, is a mouse monoclonal that detects E3 ubiquitin-protein ligase UBR1 and has been tested for use in Immunohistochemistry (Paraffin) Western Blotting.
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected E3 ubiquitin-protein ligase UBR1 in human pancreas and human kidney tissue sections.

Western Blotting Analysis: 0.5 µg/mL from a representative lot detected E3 ubiquitin-protein ligase UBR1 in C6 cell lysate.
Research Category
Signaling

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected E3 ubiquitin-protein ligase UBR1 in HEK293 cell lysate.

Target description

~200 kDa observed; 200.21 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service