Skip to Content
Merck
All Photos(1)

Key Documents

47618

Sigma-Aldrich

Fmoc-Asp(OtBu)-OH

≥98.0% (HPLC), for peptide synthesis

Synonym(s):

Fmoc-L-aspartic acid 4-tert-butyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H25NO6
CAS Number:
Molecular Weight:
411.45
Beilstein:
3635671
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26
Pricing and availability is not currently available.

Product Name

Fmoc-Asp(OtBu)-OH, ≥98.0% (HPLC)

Assay

≥98.0% (HPLC)

form

powder

optical activity

[α]20/D −24±2°, c = 1% in DMF

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

mp

148-150 °C (dec.)

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)C[C@H](NC(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O

InChI

1S/C23H25NO6/c1-23(2,3)30-20(25)12-19(21(26)27)24-22(28)29-13-18-16-10-6-4-8-14(16)15-9-5-7-11-17(15)18/h4-11,18-19H,12-13H2,1-3H3,(H,24,28)(H,26,27)/t19-/m0/s1

InChI key

FODJWPHPWBKDON-IBGZPJMESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fmoc-Asp(OtBu)-OH also known as Fmoc-L-aspartic acid 4-tert-butyl ester, is a Fmoc protected amino acid, used in solid phase peptide synthesis.

Application

Fmoc-Asp(OtBu)-OH is used to prevent the formation of aspartimide by-products in Fmoc solid phase peptide synthesis. [1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New t-butyl based aspartate protecting groups preventing aspartimide formation in Fmoc SPPS
R Behrendt, et.al.
Journal of Peptide Science, 21, 680-687 (2015)
Fengyang Xu et al.
ACS applied materials & interfaces, 8(44), 29906-29914 (2016-10-21)
In this work, we reported the generation of a novel supramolecular hydrogelator from a peptide derivative which consisted of a structural motif (e.g., Fc-FF) for supramolecular self-assembly and a functional moiety (e.g., RGD) for integrin binding. Following self-assembly in water
Jisi Zhao et al.
Molecules (Basel, Switzerland), 24(4) (2019-03-03)
Photodynamic therapy (PDT) is an established therapeutic modality for the management of cancers. Conjugation with tumor-specific small molecule ligands (e.g., short peptides or peptidomimetics) could increase the tumor targeting of PDT agents, which is very important for improving the outcome
R Ashton Lavoie et al.
Biotechnology and bioengineering, 117(2), 438-452 (2019-10-28)
The clearance of host cell proteins (HCPs) is of crucial importance in biomanufacturing, given their diversity in composition, structure, abundance, and occasional structural homology with the product. The current approach to HCP clearance in the manufacturing of monoclonal antibodies (mAbs)
Stefan Eissler et al.
Journal of peptide science : an official publication of the European Peptide Society, 23(10), 757-762 (2017-06-22)
In solid-phase peptide synthesis, the nominal batch size is calculated using the starting resin substitution and the mass of the starting resin. The starting resin substitution constitutes the basis for the calculation of a whole set of important process parameters

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service