Skip to Content
Merck
All Photos(1)

Documents

MAB2023Z

Sigma-Aldrich

Anti-Integrin β3 Antibody, clone B3A, azide free

clone B3A, Chemicon®, from mouse

Synonym(s):

CD61, MAB2023

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

B3A, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGB3(3690)

Related Categories

Specificity

Specific for human integrin beta 3 (other species not tested).

Immunogen

Human integrin alphaVbeta3 heterodimer isolated from placenta.

Application

Anti-Integrin β3 Antibody, clone B3A, azide free detects level of Integrin β3 & has been published & validated for use in FC, FUNC & WB.
Flow cytometry

Function-blocking of integrin beta3-mediated adhesion to ECM proteins.

Western blot (reducing conditions) Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Integrins

Target description

110 kDa

Physical form

Format: Purified
Protein A Purified mouse immunoglobulin in 20 mM sodium phosphate, 250 mM NaCl, pH. 7.6, with no preservatives.
Protein A purified

Storage and Stability

Maintain for 1 year at 2–8°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Positive Control: U251 or D54 human cell lines. Membrane preps are recommended for western blots.
Human endothelium cells (expression of integrin alpha V beta 3 may need to be upregulated by phorbol esters stimulation). Skin (Basement Membrane)

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

María Anguiano et al.
PloS one, 12(2), e0171417-e0171417 (2017-02-07)
Microfluidic devices are becoming mainstream tools to recapitulate in vitro the behavior of cells and tissues. In this study, we use microfluidic devices filled with hydrogels of mixed collagen-Matrigel composition to study the migration of lung cancer cells under different
Marta Irigoyen et al.
Molecular cancer, 9, 130-130 (2010-06-01)
Lung cancer is one of the most prevalent neoplasias in developed countries. Advances in patient survival have been limited and the identification of prognostic molecules is needed. Resistance to treatment is strongly related to tumor cell adhesion to the extracellular
María Anguiano et al.
PloS one, 15(1), e0220019-e0220019 (2020-01-17)
The migration of cancer cells is highly regulated by the biomechanical properties of their local microenvironment. Using 3D scaffolds of simple composition, several aspects of cancer cell mechanosensing (signal transduction, EMC remodeling, traction forces) have been separately analyzed in the
Clementina Auriemma et al.
Cellular & molecular biology letters, 15(3), 496-506 (2010-06-08)
Listeria monocytogenes enters non-phagocytic cells by binding its surface proteins inlA (internalin) and inlB to the host's E-cadherin and Met, respectively. The two internalins play either separate or cooperative roles in the colonization of infected tissues. Here, we studied bacterial
Hilary A Kenny et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 17(3), 459-471 (2010-12-15)
To understand the functional and preclinical efficacy of targeting the urokinase plasminogen activator receptor (u-PAR) in ovarian cancer. Expression of u-PAR was studied in 162 epithelial ovarian cancers, including 77 pairs of corresponding primary and metastatic tumors. The effect of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service