Skip to Content
Merck
All Photos(1)

Key Documents

F0556

Sigma-Aldrich

Human Fibronectin

from human fibroblasts, liquid, ~0.5 mg/mL, suitable for cell culture

Synonym(s):

Fibronectin

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
MDL number:
UNSPSC Code:
12352202
eCl@ss:
42020141
NACRES:
NA.75

Product Name

Fibronectin solution human fibroblasts, cell culture derived, ~0.5 mg/mL, sterile-filtered, BioReagent, suitable for cell culture

biological source

human fibroblasts

Quality Level

sterility

sterile-filtered

product line

BioReagent

form

solution

mol wt

31 kDa

packaging

pkg of 100 μL

concentration

~0.5 mg/mL

technique(s)

cell culture | mammalian: suitable

impurities

HBSAG, none detected
HCV, none detected
HIV-1/HIV-2, none detected

NCBI accession no.

UniProt accession no.

Binding Specificity

Peptide Source: Collagen

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... FN1(2335)

Looking for similar products? Visit Product Comparison Guide

General description

Fibronectin 1 is a glycoprotein of the extracellular matrix that is coded by FN1 gene. It is expressed in the plasma and at the cell surface. It is mapped to human chromosome 2q35.
The matrix protein fibronectin consists of three units that are named Fn1, Fn2 and Fn3. The plasma protein has a molar mass of 31kDa. It is present in loose connective tissue, basement membranes, and the surface of various cell types.

Application

Fibronectin solution from human fibroblasts is used for the following applications:
  • It is used in purification of human extracellular matrix (ECM)
  • Coating of tissue culture plates with extracellular matrix components
  • Used during cell culture (For xeno-free, feeder-free culture the cells were passaged in to human fibronectin along with other components)

Fibronectin from human plasma can be used with epithelial cells, mesenchymal cells, neuronal cells, fibroblasts, neural crest cells, and endothelial cells. It is recommended for use as a cell culture substratum at 1-5 μg/cm2 or 0.5-50 μg/mL. The optimal concentration does depend on cell type as well as the application and research objectives.

Biochem/physiol Actions

Fibronectin is a high molecular mass extracellular matrix glycoprotein involved in many cellular processes, including tissue repair, embryogenesis, blood clotting, cell migration/adhesion, and embryogenesis and development. It is used in vitro as a substrate to enhance adherence and proliferation of many cell types. It can bind to macromolecules such as collagen, fibrin, glycosaminoglycans, and Clq, apart from some bacteria.It serves as a critical factor in promoting cell survival and preventing programmed cell death in these cellular contexts. Fibronectin plays a significant role in the wound healing process, and in extracellular matrix (ECM) assembly hence is suitable for tissue engineering applications.

Components

Fibronectin is an adhesive glycoproteins found in both the cell surface and in plasma.

Caution

The solution should be stored at -20°C. Under these conditions, the product is stable for at least 2 years. Vortexing, excessive agitation, or additoinal freezing and thawing of reconstituted fibronectin are not recommended.

Preparation Note

This product is produced by human fibroblasts and then purified biochemically. It is supplied as a sterile solution at 0.5 mg/mL in a CAPS saline buffer. Before use, the product should be thawed slowly at 2-8°C, forming a clear solution. Avoid any vortexing or vigorous shaking of the solution, which will cause any fibronectin to “crash” out of solution. To coat plates, dilute this product in sterile HBSS solutions and coat with a minimal volume. Then, incubate at 37°C for 1-2 hours and wash 3 times with sterile HBSS and plate cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fibronectin
Fabrizio M and Angela S
Encyclopedia of Immunology, 909-913 (1998)
Amniogenesis in Human Amniotic Sac Embryoids after Exposures to Organophosphate Flame Retardants.
Xu, et al.
Environmental Health Perspectives, 131, 47007-47007 (2023)
Heidi Hakala et al.
Tissue engineering. Part A, 15(7), 1775-1785 (2009-01-10)
Long-term in vitro culture of undifferentiated human embryonic stem cells (hESCs) traditionally requires a fibroblast feeder cell layer. Using feeder cells in hESC cultures is highly laborious and limits large-scale hESC production for potential application in regenerative medicine. Replacing feeder
Patricia A Soucy et al.
Matrix biology : journal of the International Society for Matrix Biology, 28(5), 273-283 (2009-04-21)
Extracellular matrix plays a critical role in cellular development by providing signaling cues that direct morphogenesis. In order to study both the cues that natural matrix provides and endothelial cell responses to that information, human fetal lung fibroblasts were used
Priyam Banerjee et al.
Methods in molecular biology (Clifton, N.J.), 2557, 785-810 (2022-12-14)
Cancer cells utilize secretory pathways for paracrine signaling and extracellular matrix remodeling to facilitate directional cell migration, invasion, and metastasis. The Golgi apparatus is a central secretory signaling hub that is often deregulated in cancer. Here we described technologies that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service