Skip to Content
Merck
All Photos(1)

Key Documents

AP114P

Sigma-Aldrich

Goat Anti-Human IgM Antibody, Fc5µ, HRP conjugate

Chemicon®, from goat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

peroxidase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

General description

IgM constitutes about 10% of serum immunoglobulins. IgM is prominent in early immune responses to most antigens and predominates in certain antibody responses such as ′natural′ blood group antibodies. IgM (with IgD) is the major immunoglobulin expressed on the surface of B cells.

Specificity

Based on immunoelectrophoresis, the antibody reacts with the Fc5μ portion of the human IgM heavy chain. Does not react with human IgG or IgA, or with the light chains of human immunoglobulins. No reactivity against non-immunoglobulin serum proteins, but the antibody may cross-react with IgM from other species.

Immunogen

Prepared from purified human IgM Fc5μ fragment.

Application

EIA: 1:5,000-1:100,000.
Western blots: 1:5,000-1:200,000.
Immunohistochemistry: 1:500-1:5,000.

Optimal working dilutions must be determined by the end user.
Goat anti-Human IgM Antibody, Fc5μ, HRP conjugate is an antibody against Human IgM for use in ELISA & WB.
Research Category
Secondary & Control Antibodies
Research Sub Category
Fragment Specific Secondary Antibodies

Physical form

0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6 with 15mg/mL BSA. Contains no preservative. Lyophilized.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lin Sun et al.
Frontiers in bioengineering and biotechnology, 11, 1320586-1320586 (2023-12-21)
Plants are being increasingly recognized for the production of complex human proteins, including monoclonal antibodies (mAbs). Various methods have been applied to boost recombinant expression, with DNA codon usage being an important approach. Here, we transiently expressed three complex human
Cleopatra K Mugyenyi et al.
PloS one, 8(7), e68304-e68304 (2013-07-19)
Antibodies to P. falciparum apical membrane protein 1 (AMA1) may contribute to protective immunity against clinical malaria by inhibiting blood stage growth of P. falciparum, and AMA1 is a leading malaria vaccine candidate. Currently, there is limited knowledge of the
Krasimir Kostov et al.
Pathophysiology : the official journal of the International Society for Pathophysiology, 29(3), 426-434 (2022-08-24)
The increased glycation of elastin is an important factor in vascular changes in diabetes. Using the ELISA method, we determined serum levels of IgM and IgG autoantibodies to advanced glycation end products of vascular elastin (anti-AGE EL IgM and anti-AGE
Sanne Reijm et al.
Arthritis research & therapy, 23(1), 230-230 (2021-09-05)
Anti-modified protein antibodies (AMPA) targeting citrullinated, acetylated and/or carbamylated self-antigens are hallmarks of rheumatoid arthritis (RA). Although AMPA-IgG cross-reactivity to multiple post-translational modifications (PTMs) is evident, it is unknown whether the first responding B cells, expressing IgM, display similar characteristics
Krasimir Kostov et al.
Medicina (Kaunas, Lithuania), 56(5) (2020-05-15)
Background and objectives: HbA1c measurements may be useful not only in optimizing glycemic control but also as a tool for managing overall vascular risk in patients with diabetes. In the present study, we investigate the clinical significance of HbA1c as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service