Skip to Content
Merck
All Photos(1)

Documents

AP128P

Sigma-Aldrich

Goat Anti-Mouse IgM Antibody, µ chain, HRP conjugate

0.8-1.0 mg/mL, Chemicon®

Synonym(s):

Anti-Mouse IgM, Goat anti-Mouse IgM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

peroxidase conjugate

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

concentration

0.8-1.0 mg/mL

technique(s)

ELISA: suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

General description

IgM normally constitutes about 10% of serum immunoglobulins. IgM antibody is prominent in early immune responses to most antigens and predominates in certain antibody responses such as ′natural′ blood group antibodies. IgM (with IgD) is the major immunoglobulin expressed on the surface of B cells. The gene for the mu constant region contains four domains separated by short intervening sequences.

Class specific anti immunoglobulin antibodies are useful for: The characterization of malignant B cell proliferations. All but acute lymphocytic leukemias share either surface or intra cytoplasmic Ig with an isotypic restriction, which suggest the monoclonal nature of the cell population. Most of the chronic lymphocytic leukemias, non Hodgkin lymphomas and Burkitt′s lymphoma bear surface IgM, whereas plasmocytes from Waldenström′s disease bear intracytoplasmic IgM. The other isotypes are less frequently found. On the other hand multiple myelomas are usually of the IgG or IgA type.

Specificity

Based on immunoelectrophoresis, the antibody reacts with the heavy chain portion of IgM. No reactivity with IgG or non-immunoglobulin serum proteins was detected, but cross reactivity with IgM from other species may be detected.

Application

Research Category
Secondary & Control Antibodies
Research Sub Category
Fragment Specific Secondary Antibodies
This Goat anti-Mouse IgM Antibody, μ chain, HRP conjugate is validated for use in ELISA, WB for the detection of Mouse IgM.

Physical form

Freeze dried powder, made from liquid containing 0.01M phosphate buffered saline, 0.25M sodium chloride, 15 mg/mL bovine serum with no preservatives. Reconstitute in 2mL deionized water, centrifuge if not clear. Prepare a working dilution fresh for each day.
ImmunoAffinity Purified

Storage and Stability

Maintain lyophilized product at 2-8°C for up to 12 months. After reconstitution the product is stable for several weeks at 2-8°C as an undiluted liquid. For extended storage after reconstitution, add an equal volume of glycerol to make a final concentration of 50% glycerol followed by storage at -20°C in undiluted aliquots for up to 12 months. Please note the concentration of protein (and buffer salts) will decrease to one-half of the original after the addition of glycerol. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hina Singh et al.
Brain, behavior, and immunity, 118, 1-21 (2024-02-16)
Human immunodeficiency virus-1 (HIV-1) infects the central nervous system (CNS) and causes HIV-associated neurocognitive disorders (HAND) in about half of the population living with the virus despite combination anti-retroviral therapy (cART). HIV-1 activates the innate immune system, including the production
Vaccinia Virus Strain MVA Expressing a Prefusion-Stabilized SARS-CoV-2 Spike Glycoprotein Induces Robust Protection and Prevents Brain Infection in Mouse and Hamster Models.
Lorenzo, et al.
Vaccines, 11 (2023)
Andrea Largo-de la Torre et al.
Frontiers in immunology, 13, 1021556-1021556 (2022-11-08)
Livestock animals, such as swine, are an important source of Toxoplasma gondii in the human population. Currently, there is limited knowledge regarding the potential influence that the T. gondii genotype might exert on establishing infection in swine. Herein, we investigated
Nina Y Yuan et al.
Cells, 11(13) (2022-07-10)
Combination antiretroviral therapy (cART) has transformed HIV infection from a terminal disease to a manageable chronic health condition, extending patients' life expectancy to that of the general population. However, the incidence of HIV-associated neurocognitive disorders (HANDs) has persisted despite virological
Ana B Sanchez et al.
Antimicrobial agents and chemotherapy, 60(1), 168-179 (2015-10-21)
HIV-1 infection frequently causes HIV-associated neurocognitive disorders (HAND) despite combination antiretroviral therapy (cART). Evidence is accumulating that components of cART can themselves be neurotoxic upon long-term exposure. In addition, abuse of psychostimulants, such as methamphetamine, seems to aggravate HAND and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service