Skip to Content
Merck
All Photos(3)

Key Documents

AB1952P

Sigma-Aldrich

Anti-Integrin beta 1 (CD29) Antibody

CHEMICON®, rabbit polyclonal

Synonym(s):

CD29

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-Integrin β1 Antibody, cytosolic, Chemicon®, from rabbit

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rat, pig, mouse

manufacturer/tradename

Chemicon®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGB1(3688)

Specificity

Reactive with human, mouse, and rat integrin beta-1, as confirmed by western blot. Not reactive with other integrins.

Immunogen

Epitope: cytosolic
Synthetic peptide corresponding to the C-terminus of human integrin beta 1 (a.a. CTTVVNPKYEGK)

Application

Detect Integrin β1 using this Anti-Integrin β1 Antibody, cytosolic validated for use in IP & WB.
Western blot: 1-2μg/mL for detection in whole cell lysate. The antibody is effective against reduced (MW = 130kDa) and non-reduced (MW = 110kDa) proteins.

Immunoprecipitation

Optimal working dilutions must be determined by the end user.

Target description

130kDa

Physical form

Format: Purified
Protein A Purified immunoglobulin. Liquid in PBS containing 0.1% sodium azide.

Analysis Note

Control
U251 cell line Skin (Basement Membrane)

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoming Bai et al.
Scientific reports, 4, 6538-6538 (2014-10-08)
Prostaglandin E2 (PGE2) has been implicated in cell invasion in hepatocellular carcinoma (HCC), via increased β1-integrin expression and cell migration; however, the mechanism remains unclear. PGE2 exerts its effects via four subtypes of the E prostanoid receptor (EP receptor 1-4).
Jean-Baptiste Oudart et al.
Cell adhesion & migration, 15(1), 215-223 (2021-07-27)
We previously demonstrated that F4 peptide (CNPEDCLYPVSHAHQR) from collagen XIX was able to inhibit melanoma cell migrationin vitro and cancer progression in a mouse melanoma model. The aim of the present work was to study the anti-angiogenic properties of F4 peptide. We demonstrated that F4
Jinshun Pan et al.
Scientific reports, 6, 33823-33823 (2016-09-23)
Cyclooxygenase-2 (COX-2) has been implicated in cell invasion in non-small-cell lung cancer (NSCLC). However, the mechanism is unclear. The present study investigated the effect of COX-2 on β1-integrin expression and cell invasion in NSCLC. COX-2 and β1-integrin were co-expressed in
Alexia Vautrin-Glabik et al.
Frontiers in cell and developmental biology, 8, 775-775 (2020-08-28)
Angiogenesis is defined as the formation of new capillaries by sprouting from the pre-existing microvasculature. It occurs in physiological and pathological processes particularly in tumor growth and metastasis. α1, α2, α3, and α6 NC1 domains from type IV collagen were
Siyi Hu et al.
Molecular cancer, 9, 30-30 (2010-02-06)
Factors responsible for invasive and metastatic progression of prostate cancer (PCa) remain largely unknown. Previously, we reported cloning of prosaposin (PSAP) and its genomic amplification and/or overexpression in several androgen-independent metastatic PCa cell lines and lymph node metastases. PSAP is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service