Skip to Content
Merck
All Photos(1)

Key Documents

MAB2247

Sigma-Aldrich

Anti-Integrin β1 Antibody, clone 12G10

clone 12G10, Chemicon®, from mouse

Synonym(s):

Integrin beta-1, CD29, Glycoprotein Iia, GPIIA, Integrin VLA-4 subunit beta, Very late activation protein, beta polypeptide

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

12G10, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
cell culture | mammalian: suitable
flow cytometry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... ITGB1(3688)

General description

Integrin beta-1 (UniProt P05556; also known as CD29, Glycoprotein Iia, GPIIA, Integrin VLA-4 subunit beta, Very late activation protein, beta polypeptide) is encoded by the ITGB1 (also known as FNRB, MDF2, MSK12, VLA-BETA, VLAB) gene (Gene ID 3688) in human. Integrin beta-1 is a 130 kDa transmembrane glycoprotein that interact with various integrin alpha subunits (including alpha 1, alpha 2, alpha 3, alpha 4, alpha 5, and alpha 6) to form the functional receptor complexes that bind to specific extracellular matrix proteins. Integrin receptors regulate a variety of important biological functions, including embryonic development, wound repair, hemostasis, and prevention of programmed cell death. They are also implicated in abnormal pathological states such as tumor directed angiogenesis, tumor growth and metastasis. These heterodimeric receptors bridge the cytoplasmic actin cytoskeleton with proteins present in the extracellular matrix and/or on adjacent cells. Interactions between integrins and the extracellular matrix lead to the activation of signal transduction pathways and regulation of gene expression.

Specificity

MAB2247 recognizes the human beta1 integrin. This antibody has been shown to recognize a ligand-induced binding site, with antibody binding increasing in the presence of a fibronectin ligand. This antibody also enhances alpha 5 - beta 1 fibronectin interactions. MAB2247 does not cross-react with mouse or rat samples.

Immunogen

Purified human beta1 integrin preparation from HT1080 fibrosarcoma cell extract.

Application

Anti-Integrin β1 Antibody, clone 12G10 detects level of Integrin β1 & has been published & validated for use in ELISA, FC, IP, FUNC, CULT & WB.
Flow cytometry: 1:25-1:50

ELISA : 10 μg/mL

Western Blot: known to react with human beta 1 integrin subunit under non-reduced conditions only {Mould, 1995}.

Stimulatory to alpha5beta1 interaction with fibronectin {Mould, 1995}.

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Integrins

Target description

88 kDa

Physical form

Format: Purified
Protein A purified
Purified immunoglobulin. Liquid in phosphate buffer saline, with 0.1% sodium azide.

Storage and Stability

Maintain refrigerated at 2–8°C in undiluted aliquots for up one month. For long term storage maintain at -20°C for up to 12 months.

Analysis Note

Control
Tonsil, human skin tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magdalene Michael et al.
The Journal of investigative dermatology, 134(3), 773-782 (2013-09-13)
Bullous pemphigoid antigen 1 (BPAG1-e, also known as BP230) is a member of the plakin family of hemidesmosome cytoskeletal linker proteins that is encoded by an isoform of the dystonin (DST) gene. Recently, we reported two unrelated families with homozygous
Catherine T Choy et al.
Endocrine-related cancer, 21(1), 85-99 (2013-11-06)
Anosmin-1, encoded by the KAL1 gene, is an extracellular matrix (ECM)-associated protein which plays essential roles in the establishment of olfactory and GNRH neurons during early brain development. Loss-of-function mutations of KAL1 results in Kallmann syndrome with delayed puberty and
Zhenwei Lu et al.
eLife, 5 (2016-12-09)
Integrins are transmembrane receptors composed of α and β subunits. Although most integrins contain β1, canonical activation mechanisms are based on studies of the platelet integrin, αIIbβ3. Its inactive conformation is characterized by the association of the αIIb transmembrane and
Dhaarmini Rajshankar et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 26(11), 4429-4444 (2012-07-26)
IL-1β is a prominent proinflammatory cytokine that mediates degradation of extracellular matrix proteins through increased expression of matrix metalloproteinases, which involves a signaling pathway in adherent cells that is restricted by focal adhesions. Currently, the mechanism by which IL-1β affects
Julian U G Wagner et al.
Arteriosclerosis, thrombosis, and vascular biology, 38(5), 1170-1177 (2018-03-31)
Endothelial cells play important roles in tissue homeostasis and vascularization, a function that is impaired by aging. Here, we aim to decipher the role of the microenvironment underlying the impairment of endothelial cell functions by aging. RNA sequencing of isolated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service