Skip to Content
Merck
All Photos(7)

Key Documents

HPA004810

Sigma-Aldrich

Anti-IFITM1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CD225 antigen, Anti-Interferon-induced protein 17, Anti-Interferon-induced transmembrane protein 1, Anti-Interferon-inducible protein 9-27, Anti-Leu-13 antigen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

MHKEEHEVAVLGAPPSTILPRSTVINIHSETSVPDH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IFITM1(8519)

General description

IFITM1 (interferon induced transmembrane protein 1) is a member of interferon inducible transmembrane protein (IFITM) family. It is expressed in stable blood vessels of multiple organs.

Immunogen

Interferon-induced transmembrane protein 1 recombinant protein epitope signature tag (PrEST)

Application

Anti-IFITM1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

IFITM1 (interferon induced transmembrane protein 1) is associated with several cellular processes, such as homotypic cell adhesion of interfereons and cellular anti-proliferative activities. It essentially plays a role in the formation of functional blood vessels. IFITM1 regulates tight junction assembly that stabilizes EC-EC (endothelial cells) interactions during endothelial lumen formation. It acts as a biomarker in endometrial stromal and smooth muscle tumors. It performs in the antiproliferative action of IFN-γ. The suppressed expression of IFITM1 inhibits antiproliferative property of IFN-γ as well as enhances the cell growth. Studies have been suggested that IFITM1 inhibits the extracellular signaling of the kinase. By binding, it increases the transcriptional activity of p53 and stabilizes the p53 protein through inhibition of p53 phosphorylation on threonine residue.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86661

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mirella Mourad et al.
Scientific reports, 11(1), 14390-14390 (2021-07-15)
The coronavirus disease 2019 (COVID-19) pandemic affected people at all ages. Whereas pregnant women seemed to have a worse course of disease than age-matched non-pregnant women, the risk of feto-placental infection is low. Using a cohort of 66 COVID-19-positive women
Stephanie A Popson et al.
Arteriosclerosis, thrombosis, and vascular biology, 34(5), 1011-1019 (2014-03-08)
It is well established that angiogenesis is a complex and coordinated multistep process. However, there remains a lack of information about the genes that regulate individual stages of vessel formation. Here, we aimed to define the role of human interferon-induced
Hui Sun et al.
Reproductive sciences (Thousand Oaks, Calif.), 27(8), 1595-1601 (2020-05-22)
When the presence of endometriotic lesions are not evident by hematoxylin and eosin staining, CD10 is used to highlight and confirm the presence of endometriotic stroma. However, CD10 is not specific only to the endometrial stroma but is also expressed
Ji-Su Mo et al.
Immunology letters, 156(1-2), 118-122 (2013-10-15)
Interferon inducible transmembrane protein (IFITM) family genes have been implicated in several cellular processes such as the homotypic cell adhesion functions of IFNs and cellular anti-proliferative activities. We previously showed that the IFITM3 single nucleotide polymorphisms (SNPs) associated with susceptibility
Carlos E Parra-Herran et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 27(4), 569-579 (2013-09-28)
When classifying cellular uterine mesenchymal neoplasms, histological distinction of endometrial stromal from smooth muscle neoplasms can be difficult. The only widely established marker of endometrial stromal differentiation, CD10, has marginal specificity. We took a bioinformatics approach to identify more specific

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service