AB1056F
Anti-Adenovirus Antibody, FITC-conjugated
Chemicon®, from goat
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Recommended Products
biological source
goat
Quality Level
conjugate
FITC conjugate
antibody form
purified antibody
antibody product type
primary antibodies
clone
polyclonal
species reactivity
human
manufacturer/tradename
Chemicon®
technique(s)
immunofluorescence: suitable
immunohistochemistry: suitable
shipped in
wet ice
Specificity
Adenovirus, hexon assembly. No cross-reactivity with Para 1-3, Influenza A & B or RSV. Non reactive with HEp-2 cells
Immunogen
Hexon from adenovirus type 5.
Application
Anti-Adenovirus Antibody, FITC-conjugated is an antibody against Adenovirus for use in IF, IH.
Direct FA staining of target antigens in a permissive tissue culture system. Suggested dilution: 1:10-1:50. Acetone fixation of the antigen source is recommended prior to staining.
Optimal working dilutions must be determined by end user
Optimal working dilutions must be determined by end user
Research Category
Infectious Diseases
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral
Infectious Diseases - Viral
Physical form
IgG fraction conjugated to FITC. In PBS (0.01 M, pH 7.2) containing 10 mg/mL BSA and 0.1% sodium azide
Storage and Stability
Maintain at 2-8°C for 3 months or at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. Store under subdued light.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Rachael Mooney et al.
Molecular therapy oncolytics, 12, 79-92 (2019-02-06)
Oncolytic virotherapy is a promising approach for treating recurrent and/or drug-resistant ovarian cancer. However, its successful application in the clinic has been hampered by rapid immune-mediated clearance or neutralization of the virus, which reduces viral access to tumor foci. To
N Yawata et al.
Mucosal immunology, 9(1), 159-170 (2015-06-18)
The most severe form of virus-induced inflammation at the ocular surface is epidemic keratoconjunctivitis (EKC), often caused by group D human adenoviruses (HAdVs). We investigated the dynamics and mechanisms of changes in natural killer (NK) cell types in the human
Hideki Yoshida et al.
Translational oncology, 14(2), 100997-100997 (2020-12-19)
The PAX3-FOXO1 fusion gene functions as a transactivator and increases expression of many cancer-related genes. These lead to metastases and other unfavorable outcomes for alveolar rhabdomyosarcoma (ARMS) patients. In order to target ARMS with the PAX3-FOXO1 transactivator, we developed an
Gene transfer into the liver of nonhuman primates with E1-deleted recombinant adenoviral vectors: safety of readministration.
F A Nunes, E E Furth, J M Wilson, S E Raper
Human Gene Therapy null
Comparison of adenoviruses from species B, C, E, and F after intravenous delivery.
Daniel Stone, Ying Liu, Zong-Yi Li, Sebastian Tuve, Robert Strauss, Andre Lieber
Molecular Therapy null
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service