Skip to Content
Merck
All Photos(1)

Key Documents

MAB8044

Sigma-Aldrich

Anti-Adenovirus 1,2,5,6 Antibody, clone 7/48-7a

ascites fluid, clone 7/48-7a, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

clone

7/48-7a, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

isotype

IgG1

shipped in

wet ice

General description

Adenoviruses are 65-80 nm, non-enveloped, regular icosahedron pathogens often associated with respiratory and gastrointestinal illness as well as conjunctivitis. Well over 40 types of adenoviruses have currently been recognized. In addition to their deleterious effects, adenoviruses have been used in vaccine production and in gene therapy. They also have the ability to trigger cell proliferation by interfering with host cellular anti-oncogenes. Since they are easy to prepare, multiple quickly and efficiently, and have the ability to infect and deliver their genome (including researcher-created constructs) to the nucleus of a variety of post-mitotic cells, adenoviruses are extremely valuable in research and medical applications ranging from virus/host cell interaction to the insertion of novel genes or regulatory mechanisms into eukaryotic cellular systems, to delivery of therapeutic anti-cancer genes.

Specificity

Adenovirus 1,2,5,6

Immunogen

Adenovirus 1,2,5,6 hexon protein

Application

Anti-Adenovirus 1, 2, 5, 6 Antibody, clone 7/48-7a is an antibody against Adenovirus 1 for use in IF.
IFA.

Optimal working dilutions must be determined by end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Physical form

Ascites

Storage and Stability

Maintain at -20°C in convenient aliquots for up to 12 months. Do not store in a diluted format.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Infection with replication-deficient adenovirus induces changes in the dynamic instability of host cell microtubules.
Warren, JC; Rutkowski, A; Cassimeris, L
Molecular Biology of the Cell null
Rapid detection and serotyping of adenovirus by direct immunofluorescence.
Wood, S R, et al.
Journal of Medical Virology, 51, 198-201 (1997)
The contributions of microtubule stability and dynamic instability to adenovirus nuclear localization efficiency.
James C Warren,Lynne Cassimeris
Cell Motility and the Cytoskeleton null
Weidong Xu et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 22(8), 1504-1517 (2014-05-06)
We are interested in developing oncolytic adenoviruses for the treatment of prostate cancer (PCa) bone metastases. A key limitation of Adenovirus 5 (Ad5) is that upon systemic administration, it produces major liver and systemic toxicities. To address this issue, a
Natalya Kaverina et al.
Autophagy, 13(11), 1905-1923 (2017-10-06)
Formation of metastases, also known as cancer dissemination, is an important stage of breast cancer (BrCa) development. KISS1 expression is associated with inhibition of metastases development. Recently we have demonstrated that BrCa metastases to the brain exhibit low levels of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service