Skip to Content

5.08014

GR 127935 Hydrochloride

≥98% (HPLC), 5-HT1B/1D receptor antagonist, solid

Synonym(s):

GR 127935 Hydrochloride, 5-HT 1B/1D Serotonin Receptor Antagonist, GR 127935 Hydrochloride, GR-127935 hydrochloride, GR127935 hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C29H31N5O3 · xHCl
CAS Number:
Molecular Weight:
497.59 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
solid
Storage condition:
OK to freeze, protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

GR 127935 Hydrochloride,

assay

≥98% (HPLC)

Quality Segment

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

off-white

solubility

water: soluble

storage temp.

2-8°C

SMILES string

Cl.N1(CCN(CC1)c2c(ccc(c2)NC(=O)c3ccc(cc3)c4c(cc(cc4)c5n[o]c(n5)C)C)OC)C

InChI

1S/C29H31N5O3.ClH/c1-19-17-23(28-30-20(2)37-32-28)9-11-25(19)21-5-7-22(8-6-21)29(35)31-24-10-12-27(36-4)26(18-24)34-15-13-33(3)14-16-34;/h5-12,17-18H,13-16H2,1-4H3,(H,31,35);1H

InChI key

SRVVUYIJVBLEJI-UHFFFAOYSA-N

General description

A selective 5-HT1B/1D receptor antagonist. Selectivity was found to be similar for B (pki = 9.2-9.8 )and D (pki = 8.6-9.1) receptor types. GR 127935 works well in vivo and was shown to block (+)-MDMA-induced hyperactivity in rats. It alters release of serotonin in the brain, as well as reducing drug-seeking behaviour in cocaine addicted rats. In transformed rat C5 glioma cells, this inhibitor blocked 5HT with an IC50 = 97 pM.

Biochem/physiol Actions

Primary Target
5-HT1B/1D

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Rex, A. et al. 2008. Pharmacology, Biochemistry, and Behavior.88, 196.

Przegalinski, E. et al. 2008. Pharmacol. Rep.60, 798.
Bannai, M. et al. 2007. Psychopharmacol.93, 295.
McCreary A. C. et al. 1999. J. Pharmacol. Exp. Ther.290, 965.

Pauwels, P. J. et al. 1996. J. Neurochem.66, 65.
Skingle, M. et al. 1996. Behav.Brain Res.73, 157.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library