Skip to Content
Merck
All Photos(1)

Documents

8.55052

Sigma-Aldrich

Mca NovaTag® resin

Novabiochem®

Synonym(s):

N-Mca-NÆ-Fmoc-ethylenediamine MPB-AM resin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352116
NACRES:
NA.22

Quality Level

product line

NovaSyn® TG
Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

amine

storage temp.

15-25°C

General description

Mca NovaTag resin is an ideal tool for the synthesis of FRET peptide substrates based on the 7-methoxycoumarin (Mca) and 2,4-dinitrophenyl(Dnp) fluorophore/quench pair. The resin pre-loaded with the Mca fluorophore can be used directly in Fmoc SPPS. Following peptide assembly, the Dnp group can be introduced using Fmoc-Lys(Dnp)-OH, to give after TFA cleavage the desired FRET peptide substrate.

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Linkage

Replaces: 04-12-3902

Analysis Note

Color (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (Photometric determination of the Fmoc-chromophore liberated upon treatment with DBU/DMF): 0.20 - 0.50 mmol/g
Swelling Volume (in DMF): lot specific result
The polymer matrix is copoly styrene - 1 % DVB 100 - 200 mesh

Legal Information

NOVASYN is a registered trademark of Merck KGaA, Darmstadt, Germany
NOVATAG is a registered trademark of Merck KGaA, Darmstadt, Germany
Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service