Skip to Content
Merck

8.53085

Boc-N-Me-Val-OH

Novabiochem®

Synonym(s):

Boc-N-Me-Val-OH, N-α-t.-Boc-N-α-methyl-L-valine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C11H21NO4
CAS Number:
Molecular Weight:
231.29
UNSPSC Code:
12352209
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Novabiochem®

assay

≥98% (TLC)

form

powder

reaction suitability

reaction type: Boc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

mp

45-60 °C

application(s)

peptide synthesis

storage temp.

2-30°C

SMILES string

N([C@@H](C(C)C)C(=O)O)(C)C(=O)OC(C)(C)C

InChI

1S/C11H21NO4/c1-7(2)8(9(13)14)12(6)10(15)16-11(3,4)5/h7-8H,1-6H3,(H,13,14)/t8-/m0/s1

InChI key

XPUAXAVJMJDPDH-QMMMGPOBSA-N

General description

Standard building block for introduction of valine amino-acid residues by Boc SPPS.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Boc SPPS

Analysis Note

Colour (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Optical rotation α 25/D (c=1 in methanol): -95.0 - -86.0 °
Purity (TLC(011A)): ≥ 98 %
Purity (TLC(0009)): ≥ 98 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Other Notes

Replaces: 04-12-9013

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Boc-N-Me-Val-OH


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library