8.56147
Fmoc-Lys(carbamate wang resin)-AMC (100-200 mesh)
Novabiochem®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
product line
Novabiochem®
form
beads
reaction suitability
reaction type: Fmoc solid-phase peptide synthesis
manufacturer/tradename
Novabiochem®
application(s)
peptide synthesis
functional group
Fmoc
storage temp.
2-8°C
General description
A novel resin for the preparation of peptide substrates based on 3-amino-7-methylcoumarin (AMC) by Fmoc SPPS. Following Fmoc removal with 20% piperidine, peptide assembly can be effected using standard coupling methods. Treatment with 95% TFA releases the peptide-AMC directly from the solid phase.
Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Linkage
Replaces: 04-12-3917
Analysis Note
Colour (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (Photometric determination of the Fmoc-chromophore liberated upon treatment with DBU/DMF): 0.40 - 0.60 mmol/g
Swelling Volume (in DMF): lot specific result
Identity (of the substitution): passes test
The polymer matrix is copoly (styrene - 1% DVB) 100 -200 mesh. The resin is endcapped after coupling the amino acid derivative.
Appearance of substance (visual): beads
Loading (Photometric determination of the Fmoc-chromophore liberated upon treatment with DBU/DMF): 0.40 - 0.60 mmol/g
Swelling Volume (in DMF): lot specific result
Identity (of the substitution): passes test
The polymer matrix is copoly (styrene - 1% DVB) 100 -200 mesh. The resin is endcapped after coupling the amino acid derivative.
Legal Information
Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service