Skip to Content
Merck
All Photos(1)

Key Documents

8.00658

Sigma-Aldrich

Tetraethyl orthosilicate

for synthesis

Synonym(s):

Tetraethyl orthosilicate, Tetraethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)4Si
CAS Number:
Molecular Weight:
208.33
MDL number:
UNSPSC Code:
12352103
EC Index Number:
201-083-8
NACRES:
NA.22
Assay:
≥99% (GC)
bp:
168.0 °C/1013 hPa
vapor pressure:
2 hPa ( 20 °C)

vapor pressure

2 hPa ( 20 °C)

Quality Level

Assay

≥99% (GC)

form

liquid

autoignition temp.

230 °C

potency

6270 mg/kg LD50, oral (Rat)
5860 mg/kg LD50, skin (Rabbit)

expl. lim.

1.3-23 % (v/v)

pH

7 (20 °C)

bp

168.0 °C/1013 hPa

mp

-85.5 °C

transition temp

flash point 45 °C

density

0.94 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Si](OCC)(OCC)(OCC)OCC

InChI

1S/C8H20O4Si/c1-5-9-13(10-6-2,11-7-3)12-8-4/h5-8H2,1-4H3

InChI key

BOTDANWDWHJENH-UHFFFAOYSA-N

General description

Tetraethylorthosilicate (TEOS) is the ethyl ester of ortho silicic acid, which is the most readily available silicon source. It is mainly used as a crosslinking agent in silicon polymer and precursor in the preparation of silicon thin films.

Application

  • Effects of Tetraethyl Orthosilicate (TEOS) on the Light and Temperature Stability of a Pigment from Beta vulgaris and Its Potential Food Industry Applications: This study explores how TEOS can enhance the stability of natural pigments from beets, suggesting potential for improved food colorants (GA Molina et al., 2014).
  • Characterization of Chemical Vapor Deposited Tetraethyl Orthosilicate based SiO2 Films for Photonic Devices: The study details the use of TEOS in the production of silicon dioxide films for enhanced photonic device performance (J Kotcharlakota et al., 2016).
  • Comprehensive performances of carbon nanotube reinforced foam concrete with tetraethyl orthosilicate impregnation: Research indicates significant enhancements in the structural properties of foam concrete, essential for building technology (J Luo et al., 2017).
  • Mesoporous bis (trimethoxysilyl) hexane (BTMSH)/tetraethyl orthosilicate (TEOS)-based hybrid silica aerogel membranes for CO2 capture: Highlights the development of innovative aerogel membranes for effective CO2 capture, contributing to environmental management strategies (YF Lin et al., 2016).

Features and Benefits

TEOScan polymerize in alkaline, acidic, and neutral conditions, making it anadvantageous precursor

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.932 - 0.934
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

113.0 °F - closed cup

Flash Point(C)

45 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Real-time study of CVD growth of silicon oxide on rutile TiO2 (110) using tetraethyl orthosilicate
Chaudhary S, et al.
The Journal of Physical Chemistry C, 119(33), 19149-19161 (2015)
Effects of tetraethyl orthosilicate (TEOS) on the light and temperature stability of a pigment from Beta vulgaris and its potential food industry applications
Molina GA, et al.
Molecules (Basel), 19(11), 17985-18002 (2014)
Catalytic Cracking of n-Dodecane to Chemicals: Effect of Variable-Morphological ZSM-5 Zeolites Synthesized Using Various Silica Sources
Sanhoob MA, et al.
ACS Omega, 7(12), 10317-10329 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service