Skip to Content
Merck
All Photos(1)

Key Documents

567310-M

Millipore

SKF-96365, Hydrochloride

Inhibits receptor-mediated Ca2+ entry in activated platelets, neutrophils, and endothelial cells at concentrations that do not affect internal Ca2+ release.

Synonym(s):

SKF-96365, Hydrochloride, 1-[β-[3-(4-Methoxyphenyl)propoxy]-4-methoxyphenethyl]-1H-imidazole, HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H26N2O3 · xHCl
CAS Number:
Molecular Weight:
366.45 (free base basis)
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 2 mg/mL

storage temp.

10-30°C

SMILES string

[Cl-].[n+H]1c[n](cc1)CC(OCCCc3ccc(cc3)OC)c2ccc(cc2)OC

InChI

1S/C22H26N2O3.ClH/c1-25-20-9-5-18(6-10-20)4-3-15-27-22(16-24-14-13-23-17-24)19-7-11-21(26-2)12-8-19;/h5-14,17,22H,3-4,15-16H2,1-2H3;1H

InChI key

FWLPKVQUECFKSW-UHFFFAOYSA-N

General description

Inhibits receptor-mediated Ca2+ entry in stimulated platelets (IC50 = 8-12 µM in intact platelets), neutrophils, and endothelial cells (IC50 = 30 µM for human endothelial cells) at concentrations that do not affect internal Ca2+ release. Also inhibits voltage-gated Ca2+ entry in excitable cells. Inhibits the histamine-induced formation of nitric oxide in human endothelial cells.

Biochem/physiol Actions

Primary Target
Inhibits receptor-mediated Ca2+ entry

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Leung, Y.-M., et al. 1996. Biochem. Pharmacol.51, 605.
Schwartz, G., et al. 1994. Cell Calcium15, 45.
Cabello, O.A. and Schilling, W.P. 1993. Biochem. J. 295, 357.
Graier, W.F., et al. 1992. Biochem. Biophys. Res. Commun. 186, 1539.
Blayney, L.M., et al. 1991. Biochem. J. 273, 803.
Fasolato, C., et al. 1990. J. Biol. Chem. 265, 20351.
Merritt, J.E., et al. 1990. Biochem. J.271, 515.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service