Skip to Content
Merck
All Photos(1)

Key Documents

S7809

Sigma-Aldrich

SKF-96365

≥98% (HPLC), solid

Synonym(s):

1-[β-(3-(4-Methoxyphenyl)propoxy)-4-methoxyphenethyl]-1H-imidazole hydrochloride, 1-[2-(4-Methoxyphenyl)-2-[3-(4-methoxyphenyl)propoxy]ethyl]imidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H26N2O3 · HCl
CAS Number:
Molecular Weight:
402.91
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

color

white

solubility

DMSO: >20 mg/mL
H2O: >20 mg/mL

SMILES string

Cl.COc1ccc(CCCOC(Cn2ccnc2)c3ccc(OC)cc3)cc1

InChI

1S/C22H26N2O3.ClH/c1-25-20-9-5-18(6-10-20)4-3-15-27-22(16-24-14-13-23-17-24)19-7-11-21(26-2)12-8-19;/h5-14,17,22H,3-4,15-16H2,1-2H3;1H

InChI key

FWLPKVQUECFKSW-UHFFFAOYSA-N

Application

SKF-96365 has been used as :
  • a nonselective transient receptor potential 6 (TRPC6) blocker in human podocytes
  • an antagonist of store-operated Ca2+ influx in neurons
  • an inhibitor for transient receptor potential 3 (TRPC3) in mice

Biochem/physiol Actions

SKF-96365 with an alkylated imidazole ring is a selective inhibitor of receptor-mediated Ca2+ entry and voltage-gated Ca2+ entry . SKF-96365 mediated depolarization of smooth cardiac muscle membrane and had no impact on the acetylcholine (ACh)-induced depolarization. It is an inhibitor for SOCE (store-operated calcium entry). It also inhibits Homer1 protein expression. SKF-96365 elicits protective functionality in 1-methyl-4-phenylpyridinium(MPP) mediated cytotoxicity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protective effects of SKF-96365, a non-specific inhibitor of SOCE, against MPP+-induced cytotoxicity in PC12 cells: potential role of Homer1
Chen T, et al.
PLoS ONE, 8(1), e55601-e55601 (2013)
Effects of inhibitors of nonselective cation channels on the acetylcholine-induced depolarization of circular smooth muscle from the guinea-pig stomach antrum
Hotta A, et al.
Journal of Smooth Muscle Research = Nihon Heikatsukin Gakkai Kikanshi, 41(6), 313-327 (2005)
J E Merritt et al.
The Biochemical journal, 271(2), 515-522 (1990-10-15)
A novel inhibitor of receptor-mediated calcium entry (RMCE) is described. SK&F 96365 (1-(beta-[3-(4-methoxy-phenyl)propoxy]-4-methoxyphenethyl)-1H- imidazole hydrochloride) is structurally distinct from the known 'calcium antagonists' and shows selectivity in blocking RMCE compared with receptor-mediated internal Ca2+ release. Human platelets, neutrophils and endothelial
Arnaud Guilbert et al.
BMC cancer, 8, 125-125 (2008-05-03)
TRP channels have been shown to be involved in tumour generation and malignant growth. However, the expression of these channels in breast cancer remains unclear. Here we studied the expression and function of endogenous TRPC6 channels in a breast cancer
Marc Anderson et al.
Journal of cellular physiology, 229(4), 434-442 (2013-09-17)
Angiotensin II (AII) plays a major role in the progression of chronic kidney diseases. Podocytes are essential components of the ultrafiltration apparatus, and are targets for AII signaling. AII has been shown to increase generation of reactive oxygen species (ROS)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service