Skip to Content
Merck
All Photos(1)

Key Documents

365253

Sigma-Aldrich

Gö 6976

InSolution, ≥95%, Inhibitor of protein kinase C

Synonym(s):

InSolution Gö 6976, Go 6976, Solution, 12-(2-Cyanoethyl)-6,7,12,13-tetrahydro-13-methyl-5-oxo-5H-indolo(2,3-a)pyrrolo(3,4-c)-carbazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H18N4O
CAS Number:
Molecular Weight:
378.43
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

solubility

DMSO: 5 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

[n]1(c2c3[n](c4c(c3c5c(c2c6c1cccc6)CNC5=O)cccc4)C)CCC#N

InChI

1S/C24H18N4O/c1-27-17-9-4-2-7-14(17)20-21-16(13-26-24(21)29)19-15-8-3-5-10-18(15)28(12-6-11-25)23(19)22(20)27/h2-5,7-10H,6,12-13H2,1H3,(H,26,29)

InChI key

VWVYILCFSYNJHF-UHFFFAOYSA-N

General description

Inhibitor of protein kinase C (PKC; IC50 = 7.9 nM for rat brain). Selectively inhibits Ca2+-dependent PKCα-isozyme (IC50 = 2.3 nM) and PKCβI (IC50 = 6.2 nM). Does not affect the kinase activity of the Ca2+-independent PKC δ-, ε-, and ζ-isoenzymes even at micromolar levels. A potent antagonist of HIV-1 induction.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PKC
Product competes with ATP.
Reversible: yes
Target IC50: 7.9 nM, 2.3 nM, and 6.2 nM, for rat brain PKC, Ca2+-dependent PKC α-isozyme, and PKCβI, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 500 µg/ml solution of Gö 6976 (Cat. No. 365250) in anhydrous DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Gschwendt, M., et al. 1996. FEBS Lett.392, 77.
Wenzel-Seifert, K., et al. 1994. Biochem. Biophys. Res. Commun.200, 1536.
Martiny-Baron, G., et al. 1993. J. Biol. Chem.268, 9194.
Qatsha, K.A., et al. 1993. Proc. Natl. Acad. Sci. USA90, 674.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup - (Dimethylsulfoxide)

Flash Point(C)

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li-Mei Chen et al.
Bioscience reports, 41(7) (2021-07-10)
The serine protease prostasin is a negative regulator of lipopolysaccharide-induced inflammation and has a role in the regulation of cellular immunity. Prostasin expression in cancer cells inhibits migration and metastasis, and reduces epithelial-mesenchymal transition. Programmed death-ligand 1 (PD-L1) is a
An-Angela N Van et al.
The Journal of biological chemistry, 296, 100445-100445 (2021-02-23)
Within the AGC kinase superfamily, gene fusions resulting from chromosomal rearrangements have been most frequently described for protein kinase C (PKC), with gene fragments encoding either the C-terminal catalytic domain or the N-terminal regulatory moiety fused to other genes. Kinase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service