Skip to Content
Merck
All Photos(1)

Documents

236020

Sigma-Aldrich

Etomoxir

≥98% (HPLC), solid, CPT-1 inhibitor

Synonym(s):

Etomoxir, R(+)-2-[6-(4-Chlorophenoxy)hexyl]-oxirane-2-carboxylic acid sodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H18ClO4 · Na · H2O
CAS Number:
Molecular Weight:
338.76
UNSPSC Code:
12352200
NACRES:
NA.77

product name

Etomoxir, A cell-permeable, irreversible, and stereospecific compound that is shown to inhibit carnitine palmitoyltransferase (CPT)-1 and DGAT activity in the mitochondria of rat heart H9c2 myoblastic cells.

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL (or water)

shipped in

wet ice

storage temp.

2-8°C

InChI

1S/C15H19ClO4.Na/c16-12-5-7-13(8-6-12)19-10-4-2-1-3-9-15(11-20-15)14(17)18;/h5-8H,1-4,9-11H2,(H,17,18);/q;+1/p-1/t15-;/m1./s1

InChI key

RPACBEVZENYWOL-XFULWGLBSA-M

Warning

Toxicity: Toxic (F)

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria A Ahonen et al.
Molecular medicine (Cambridge, Mass.), 28(1), 68-68 (2022-06-18)
Thyroid hormone responsive protein (THRSP) is a lipogenic nuclear protein that is highly expressed in murine adipose tissue, but its role in humans remains unknown. We characterized the insulin regulation of THRSP in vivo in human adipose tissue biopsies and
Mengqi Li et al.
Cell research (2024-06-20)
The shift of carbon utilization from primarily glucose to other nutrients is a fundamental metabolic adaptation to cope with decreased blood glucose levels and the consequent decline in glucose oxidation. AMP-activated protein kinase (AMPK) plays crucial roles in this metabolic
Aude Angelini et al.
STAR protocols, 3(2), 101392-101392 (2022-05-24)
Metabolic switches play a critical role in the pathophysiology of cardiac diseases, including heart failure. Here, we describe an assay for long-chain fatty acid oxidation in neonatal mouse cardiomyocytes by using a SeaHorse Flux Analyzer (Agilent). This protocol is a
Kristina Seiler et al.
Cell death & disease, 13(5), 448-448 (2022-05-11)
The family of hexokinases (HKs) catalyzes the first step of glycolysis, the ATP-dependent phosphorylation of glucose to glucose-6-phosphate. While HK1 and HK2 are ubiquitously expressed, the less well-studied HK3 is primarily expressed in hematopoietic cells and tissues and is highly
Albert Pérez-Martí et al.
eLife, 11 (2022-05-14)
In diabetic patients, dyslipidemia frequently contributes to organ damage such as diabetic kidney disease (DKD). Dyslipidemia is associated with both excessive deposition of triacylglycerol (TAG) in lipid droplets (LDs) and lipotoxicity. Yet, it is unclear how these two effects correlate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service