230734
Chorionic Gonadotropin, Human Urine, Standard Grade
Chorionic Gonadotropin, Human Urine, Standard Grade, CAS 9002-61-3, is a native glycoprotein hormone that is synthesized by chorionic tissue of the placenta and found in urine during pregnancy.
Synonym(s):
Chorionic Gonadotropin, Human Urine, Standard Grade, hCG
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
description
Protein Concentration/ Report Result, weight
sterility
non-sterile
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
color
off-white
pI
4.9
solubility
water: soluble
shipped in
ambient
storage temp.
−20°C
General description
Native, human chorionic gonadotropin from urine. Glycoprotein hormone synthesized by chorionic tissue of the placenta and found in urine during pregnancy. Also present in body fluids of patients with trophoblastic disease and ovarian tumors.
Native, human chorionic gonadotropin from urine. Glycoprotein hormone synthesized by chorionic tissue of the placenta and found in urine during pregnancy. Also present in body fluids of patients with trophoblastic disease and ovarian tumors. ≥3000 IU/mg (Who 3rd IS 75/537).
Biochem/physiol Actions
≥3000 IU/mg solids (Who 3rd IS 75/537)
Warning
Toxicity: Carcinogenic / Teratogenic (D)
Reconstitution
Following reconstitution, aliquot and freeze (-20°C). Avoid freeze/thaw cycles of reconstituted solutions. Stock solutions are stable for up to 3 months at -20°C.
Other Notes
Chandy, K.G., et al. 1990. Science247, 973.
Smith, D.B. and Johnson, K.S. 1988. Gene67, 31.
Tempel, B.L., et al. 1988. Nature332, 837.
Smith, D.B. and Johnson, K.S. 1988. Gene67, 31.
Tempel, B.L., et al. 1988. Nature332, 837.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Repr. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Kimberly Mitchell et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(23), 12772-12783 (2020-05-30)
The luteinizing hormone surge is essential for fertility as it triggers ovulation in females and sperm release in males. We previously reported that secretoneurin-a, a neuropeptide derived from the processing of secretogranin-2a (Scg2a), stimulates luteinizing hormone release, suggesting a role
Xuzhe Zhang et al.
Molecular biology and evolution, 37(2), 507-523 (2019-10-22)
Evolution of highly invasive placentation in the stem lineage of eutherians and subsequent extension of pregnancy set eutherians apart from other mammals, that is, marsupials with short-lived placentas, and oviparous monotremes. Recent studies suggest that eutherian implantation evolved from marsupial
Elisabet Rosàs-Canyelles et al.
Science advances, 6(17), eaay1751-eaay1751 (2020-06-05)
The process by which a zygote develops from a single cell into a multicellular organism is poorly understood. Advances are hindered by detection specificity and sensitivity limitations of single-cell protein tools and by challenges in integrating multimodal data. We introduce
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service